Gene Transcripts

Gene Symbol
Lrrc3b
Entrez Gene ID
218763
Gene Name
leucine rich repeat containing 3B
Gene Aliases
BC019794, LRP15
Chromosome Location
Chr. 14: 15357515 - 15438987 on Build GRCm38
UniGene ID
Mm.138039
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146052.4 NP_666164.1 2 713
GenBank mRNA AK043406.1 - 2 600
BC019794.1 - 2 475 4481142

View Full Product Details