Gene Transcripts

Gene Symbol
St6gal2
Entrez Gene ID
240119
Gene Name
beta galactoside alpha 2,6 sialyltransferase 2
Gene Aliases
C230064G14Rik, St6galII, mKIAA1877
Chromosome Location
Chr. 17: 55444358 - 55514581 on Build GRCm38
UniGene ID
Mm.311687
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172829.2 NP_766417.1 5 1508
GenBank mRNA AK082566.1 - 5 1509
BC125586.1 - 4 1378 40130101
BC138142.1 - 4 1378 8861161

View Full Product Details