Gene Transcripts
- Gene Symbol
- NCF2
- Entrez Gene ID
- 4688
- Gene Name
- neutrophil cytosolic factor 2
- Gene Aliases
- NCF-2, NOXA2, P67-PHOX, P67PHOX
- Chromosome Location
- Chr. 1: 183555562 - 183590921 on Build GRCh38
- UniGene ID
- Hs.587558
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000433.3 |
NP_000424.2
|
11, 12 |
1305 |
|
NM_001127651.2 |
NP_001121123.1
|
12, 13 |
1142 |
|
NM_001190789.1 |
NP_001177718.1
|
9, 10 |
1062 |
|
NM_001190794.1 |
NP_001177723.1
|
10, 11 |
1170 |
|
XM_005245207.1 |
XP_005245264.1
|
10, 11 |
1126 |
|
XM_011509580.1 |
XP_011507882.1
|
12, 13 |
1143 |
|
XM_011509581.1 |
XP_011507883.1
|
12, 13 |
1130 |
|
GenBank mRNA |
AB209647.1 |
- |
9, 10 |
4575 |
|
AF527950.1 |
- |
11, 12 |
1052 |
|
AK296672.1 |
- |
9, 10 |
918 |
|
AK298713.1 |
- |
10, 11 |
1079 |
|
AK312666.1 |
- |
11, 12 |
1215 |
|
BC001606.1 |
- |
11, 12 |
1282 |
3542841 |
BT007439.1 |
- |
11, 12 |
1030 |
|
KU178216.1 |
- |
11, 12 |
1030 |
|
M32011.1 |
- |
12, 13 |
1097 |
|
View Full Product Details