Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000940.2 | NP_000931.1 | 4 | 253 | |
GenBank mRNA | AF320003.1 | - | 4 | 228 | |
AF329433.1 | - | 4 | 228 | ||
AK304282.1 | - | 5 | 390 | ||
AV655538.1 | - | 4 | 253 | ||
BC070374.1 | - | 5 | 239 | 4710338 | |
CD014095.1 | - | 5 | 385 | ||
CD014097.1 | - | 4 | 241 | ||
CD014100.1 | - | 4 | 241 | ||
CD014102.1 | - | 4 | 241 | ||
L48516.1 | - | 4 | 189 |