Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001131027.1 | NP_001124499.1 | 3 | 389 | |
NM_003687.3 | NP_003678.2 | 3 | 389 | ||
GenBank mRNA | AF153882.1 | - | 3 | 353 | |
AF154335.1 | - | 3 | 376 | ||
AK225407.1 | - | 3 | 351 | ||
AK313508.1 | - | 3 | 354 | ||
BC003096.1 | - | 3 | 340 | 3501794 | |
BC016765.1 | - | 3 | 391 | 4132234 | |
BT007019.1 | - | 3 | 312 | ||
DN997097.1 | - | 3 | 408 | ||
KU178433.1 | - | 3 | 312 | ||
KU178434.1 | - | 3 | 312 | ||
X93510.1 | - | 3 | 350 |