Gene Transcripts
- Gene Symbol
- TENM1
- Entrez Gene ID
- 10178
- Gene Name
- teneurin transmembrane protein 1
- Gene Aliases
- ODZ1, ODZ3, TEN-M1, TNM, TNM1
- Chromosome Location
- Chr. X: 124374680 - 125204305 on Build GRCh38
- UniGene ID
- Hs.23796
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163278.1 |
NP_001156750.1
|
5, 6 |
1089 |
|
NM_014253.3 |
NP_055068.2
|
5, 6 |
1089 |
|
XM_011531230.2 |
XP_011529532.1
|
8, 9 |
1730 |
|
XM_017029208.1 |
XP_016884697.1
|
8, 9 |
1730 |
|
XM_017029209.1 |
XP_016884698.1
|
9, 10 |
1785 |
|
XM_017029210.1 |
XP_016884699.1
|
7, 8 |
1658 |
|
XM_017029211.1 |
XP_016884700.1
|
8, 9 |
1736 |
|
XM_017029212.1 |
XP_016884701.1
|
8, 9 |
1347 |
|
XM_017029213.1 |
XP_016884702.1
|
7, 8 |
1659 |
|
XM_017029214.1 |
XP_016884703.1
|
8, 9 |
1730 |
|
XM_017029215.1 |
XP_016884704.1
|
8, 9 |
1729 |
|
XM_017029216.1 |
XP_016884705.1
|
8, 9 |
1729 |
|
GenBank mRNA |
AF100772.1 |
- |
5, 6 |
1089 |
|
BC140783.1 |
- |
5, 6 |
1086 |
9021651 |
View Full Product Details