Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015439.2 | NP_056254.1 | 3 | 665 | |
XM_011535728.2 | XP_011534030.1 | 3 | 668 | ||
GenBank mRNA | AJ420568.1 | - | 3 | 416 | |
AK300672.1 | - | 3 | 416 | ||
AL050197.1 | - | 3 | 415 | ||
AY167571.1 | - | 3 | 683 | ||
BC004464.1 | - | 3 | 400 | 2820737 | |
BC013019.2 | - | 3 | 397 | ||
BC080566.1 | - | 3 | 399 | ||
BC108717.1 | - | 3 | 588 | 6068596 |