Gene Transcripts

Gene Symbol
YPEL5
Entrez Gene ID
51646
Gene Name
yippee like 5
Gene Aliases
CGI-127
Chromosome Location
Chr. 2: 30146568 - 30160533 on Build GRCh38
UniGene ID
Hs.515890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127399.1 NP_001120871.1 4 437
NM_001127400.1 NP_001120872.1 4 673
NM_001127401.1 NP_001120873.1 5 734
NM_016061.2 NP_057145.1 3 376
XM_017004318.1 XP_016859807.1 5 972
XM_017004319.1 XP_016859808.1 4 410
XM_017004320.1 XP_016859809.1 4 911
XM_017004321.1 XP_016859810.1 4 603
XM_017004322.1 XP_016859811.1 4 834
GenBank mRNA AB098739.1 - 2 173
AF135161.1 - 3 280
AF151885.1 - 3 298
AF172940.1 - 3 300
AK025730.1 - 4 314
AK307099.1 - 2 89
BC000836.1 - 4 251 3456031
BC047237.1 - 4 834 5269563

View Full Product Details