Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001131062.1 | NP_001124534.1 | 3 | 961 | |
NM_001131063.1 | NP_001124535.1 | 3 | 946 | ||
NM_024632.5 | NP_078908.1 | 4 | 1084 | ||
NR_024084.1 | 5 | 1128 | |||
GenBank mRNA | AF085957.1 | - | 2 | 436 | |
AI199517.1 | - | 3 | 358 | ||
AI436556.1 | - | 3 | 373 | ||
AK021588.1 | - | 4 | 1084 | ||
AY341060.1 | - | 4 | 1027 | ||
AY846876.1 | - | 4 | 436 | ||
BC009829.2 | - | 4 | 1018 | 3937610 |