Gene Transcripts

Gene Symbol
PIP5KL1
Entrez Gene ID
138429
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Gene Aliases
PIPKH, bA203J24.5
Chromosome Location
Chr. 9: 127920886 - 127931958 on Build GRCh38
UniGene ID
Hs.734928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135219.1 NP_001128691.1 8 781
NM_173492.1 NP_775763.1 3 337
GenBank mRNA BC042184.1 - 3 337

View Full Product Details