Gene Transcripts

Gene Symbol
FMC1
Entrez Gene ID
154791
Gene Name
formation of mitochondrial complex V assembly factor 1 homolog
Gene Aliases
C7orf55, HSPC268
Chromosome Location
Chr. 7: 139340450 - 139346319 on Build GRCh38
UniGene ID
Hs.718441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197964.4 NP_932068.2 2 421
NR_073058.1 3 294
NR_073059.1 2 200
NR_073060.1 2 180
GenBank mRNA AA620996.1 - 1 263
AF161386.1 - 2 227
BC008467.1 - 2 224 4276817
BI117784.1 - 2 421
BM672187.1 - 1 278
BU852633.1 - 2 204
CB125801.1 - 2 402

Gene Symbol
C7orf55-LUC7L2
Entrez Gene ID
100996928
Gene Name
C7orf55-LUC7L2 readthrough
Gene Aliases
Chromosome Location
Chr. 7: 139341132 - 139423457 on Build GRCh38
UniGene ID
Hs.731488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244584.2 NP_001231513.1 2 421
GenBank mRNA AK297642.1 - 2 193

View Full Product Details