Gene Transcripts

Gene Symbol
CNTN3
Entrez Gene ID
5067
Gene Name
contactin 3
Gene Aliases
BIG-1, PANG, PCS
Chromosome Location
Chr. 3: 74262568 - 74614448 on Build GRCh38
UniGene ID
Hs.12723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020872.2 NP_065923.1 5 509
XM_005264757.3 XP_005264814.1 6 619
XM_011533768.2 XP_011532070.1 5 4141
XM_017006507.1 XP_016861996.1 6 1189
XM_017006508.1 XP_016861997.1 5 414
GenBank mRNA AB040929.1 - 3 158
BC150608.1 - 5 509 9056978
DR002512.1 - 5 508

View Full Product Details