Gene Transcripts

Gene Symbol
PTBP1
Entrez Gene ID
5725
Gene Name
polypyrimidine tract binding protein 1
Gene Aliases
HNRNP-I, HNRNPI, HNRPI, PTB, PTB-1, PTB-T, PTB2, PTB3, PTB4, pPTB
Chromosome Location
Chr. 19: 797392 - 812327 on Build GRCh38
UniGene ID
Hs.172550
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002819.4 NP_002810.1 15 1708
NM_031990.3 NP_114367.1 15 1687
NM_031991.3 NP_114368.1 14 1630
XM_005259597.2 XP_005259654.1 15 2211
XM_005259598.2 XP_005259655.1 14 2133
GenBank mRNA AB208910.1 - 12 4108
BC002397.2 - 15 1675 2821531
BC004383.1 - 14 1540 3615251
BC013694.1 - 15 1621 3863892
BC023219.1 - 15 1674
BT006819.1 - 14 1602
X60648.1 - 14 1552
X62006.1 - 14 1569
X65371.1 - 15 1626
X65372.1 - 15 1647
X66975.1 - 15 1689

View Full Product Details