Gene Transcripts

Gene Symbol
ATP9A
Entrez Gene ID
10079
Gene Name
ATPase phospholipid transporting 9A (putative)
Gene Aliases
ATPIIA
Chromosome Location
Chr. 20: 51596775 - 51768411 on Build GRCh38
UniGene ID
Hs.649234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006045.2 NP_006036.1 2, 3 264
XM_011528481.2 XP_011526783.1 2, 3 292
GenBank mRNA AB014511.1 - 2, 3 181
AK299064.1 - 2, 3 193

View Full Product Details