Gene Transcripts

Gene Symbol
ODAM
Entrez Gene ID
54959
Gene Name
odontogenic, ameloblast asssociated
Gene Aliases
APIN
Chromosome Location
Chr. 4: 70195728 - 70204576 on Build GRCh38
UniGene ID
Hs.143811
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017855.3 NP_060325.3 9 790
XM_005265698.3 XP_005265755.1 10 854
XM_006714245.3 XP_006714308.1 9 783
XM_017008331.1 XP_016863820.1 8 746
GenBank mRNA AK000520.1 - 6 441
AK098169.1 - 2 484
AK225547.1 - 6 421
BC017796.1 - 5 397 4689080
EF113908.1 - 9 792

View Full Product Details