Gene Transcripts

Gene Symbol
RAD51AP2
Entrez Gene ID
729475
Gene Name
RAD51 associated protein 2
Gene Aliases
Chromosome Location
Chr. 2: 17510584 - 17530414 on Build GRCh38
UniGene ID
Hs.515802
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099218.2 NP_001092688.1 1 2457
NM_001321233.1 NP_001308162.1 5 2815
XM_011533084.1 XP_011531386.1 1 2457
GenBank mRNA AK310498.1 - 6 3062
DQ860102.1 - 1 2457

View Full Product Details