Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145646.1 | NP_001139118.1 | 2 | 281 | |
NM_031301.3 | NP_112591.2 | 2 | 281 | ||
XM_011522105.2 | XP_011520407.1 | 2 | 312 | ||
GenBank mRNA | AB189172.1 | - | 2 | 221 | |
AB197122.1 | - | 3 | 285 | ||
AF508794.1 | - | 2 | 221 | ||
AK291204.1 | - | 2 | 223 | ||
AK307456.1 | - | 1 | 29 | ||
AK307725.1 | - | 2 | 343 | ||
AL136671.1 | - | 2 | 227 | ||
AY358698.1 | - | 2 | 215 | ||
BC020905.1 | - | 2 | 223 | 4720647 | |
BI825203.1 | - | 2 | 277 | ||
DA397427.1 | - | 2 | 281 |