Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042533.2 | NP_001035998.1 | 8 | 1669 | |
NM_001261829.1 | NP_001248758.1 | 8 | 1199 | ||
NM_032778.5 | NP_116167.3 | 8 | 1666 | ||
NM_153182.3 | NP_694822.2 | 8 | 1202 | ||
XM_005247838.4 | XP_005247895.1 | 8 | 1419 | ||
XM_011513244.2 | XP_011511546.1 | 4 | 931 | ||
GenBank mRNA | AB083189.1 | - | 8 | 1305 | |
AB083190.1 | - | 8 | 1302 | ||
AK027299.1 | - | 8 | 1149 | ||
AY302110.3 | - | 8 | 1372 | ||
AY390536.1 | - | 8 | 1354 | ||
AY456380.1 | - | 8 | 936 | ||
BC014928.1 | - | 8 | 1356 | 3916148 | |
CR627479.1 | - | 7 | 955 | ||
DQ453796.1 | - | 8 | 1166 |