Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68905991-68906396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGHMBP2
Entrez Gene ID
3508
Gene Name
immunoglobulin mu binding protein 2
Gene Aliases
CATF1, CMT2S, HCSA, HMN6, SMARD1, SMUBP2, ZFAND7
UniGene
Hs.503048
Gene Chromosome Location
Chr.11: 68903851-68940602 on Build GRCh38
Transcripts
NM_002180.2 XM_005273974.2 XM_005273976.1 XM_017017669.1 XM_017017671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3870032 c.235C>T p.P79S Chr.11: 68906217-68906217 on Build GRCh38
690271 c.128G>T p.R43L Chr.11: 68906110-68906110 on Build GRCh38
7241283 c.185G>T p.R62L Chr.11: 68906167-68906167 on Build GRCh38
6347330 c.161G>T p.S54I Chr.11: 68906143-68906143 on Build GRCh38
1605142 c.208A>T p.R70W Chr.11: 68906190-68906190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144361739 Chr.11: 68906125-68906125 on Build GRCh38
rs777016529 Chr.11: 68906288-68906288 on Build GRCh38
rs377313089 Chr.11: 68906289-68906289 on Build GRCh38
rs745812844 Chr.11: 68906236-68906236 on Build GRCh38
rs199586231 Chr.11: 68906085-68906085 on Build GRCh38
rs1021399630 Chr.11: 68906110-68906110 on Build GRCh38
rs34617762 Chr.11: 68906162-68906162 on Build GRCh38
rs373149890 Chr.11: 68906033-68906033 on Build GRCh38
rs111780234 Chr.11: 68906290-68906290 on Build GRCh38
rs749258691 Chr.11: 68906077-68906079 on Build GRCh38
rs987807139 Chr.11: 68906278-68906278 on Build GRCh38
rs748284174 Chr.11: 68906249-68906249 on Build GRCh38
rs940040078 Chr.11: 68906161-68906161 on Build GRCh38
rs368706911 Chr.11: 68906111-68906111 on Build GRCh38
rs768186724 Chr.11: 68906059-68906059 on Build GRCh38
rs372000714 Chr.11: 68906120-68906120 on Build GRCh38
rs535394484 Chr.11: 68906263-68906263 on Build GRCh38
rs762518487 Chr.11: 68906302-68906302 on Build GRCh38
rs201649839 Chr.11: 68906148-68906148 on Build GRCh38
rs562446187 Chr.11: 68906060-68906060 on Build GRCh38
rs371158900 Chr.11: 68906058-68906058 on Build GRCh38
rs770769976 Chr.11: 68906021-68906021 on Build GRCh38
rs982646623 Chr.11: 68906118-68906118 on Build GRCh38
rs118015540 Chr.11: 68906247-68906247 on Build GRCh38
rs760649755 Chr.11: 68906246-68906246 on Build GRCh38
rs747736432 Chr.11: 68906061-68906061 on Build GRCh38
rs773543257 Chr.11: 68906193-68906193 on Build GRCh38
rs201488313 Chr.11: 68906265-68906265 on Build GRCh38
rs375646306 Chr.11: 68906055-68906055 on Build GRCh38
rs137852668 Chr.11: 68906103-68906103 on Build GRCh38
rs139237340 Chr.11: 68906204-68906204 on Build GRCh38
rs774440957 Chr.11: 68906115-68906115 on Build GRCh38
rs747822666 Chr.11: 68906144-68906144 on Build GRCh38
rs188719450 Chr.11: 68906189-68906189 on Build GRCh38
rs2228206 Chr.11: 68906205-68906205 on Build GRCh38
rs757006949 Chr.11: 68906224-68906224 on Build GRCh38
rs373831746 Chr.11: 68906051-68906051 on Build GRCh38
rs935835880 Chr.11: 68906296-68906296 on Build GRCh38
rs756513980 Chr.11: 68906132-68906132 on Build GRCh38
rs200089714 Chr.11: 68906109-68906109 on Build GRCh38
rs771226710 Chr.11: 68906271-68906271 on Build GRCh38
rs771872053 Chr.11: 68906068-68906068 on Build GRCh38
rs772945179 Chr.11: 68906071-68906071 on Build GRCh38
rs920864493 Chr.11: 68906175-68906175 on Build GRCh38
rs192847221 Chr.11: 68906305-68906305 on Build GRCh38
rs112581114 Chr.11: 68906344-68906344 on Build GRCh38
rs563922049 Chr.11: 68906190-68906190 on Build GRCh38
rs779643841 Chr.11: 68906056-68906056 on Build GRCh38
rs755468547 Chr.11: 68906198-68906198 on Build GRCh38
rs201692151 Chr.11: 68906147-68906147 on Build GRCh38
rs766757770 Chr.11: 68906194-68906194 on Build GRCh38
rs776499527 Chr.11: 68906159-68906159 on Build GRCh38
rs571887680 Chr.11: 68906149-68906149 on Build GRCh38
rs762291036 Chr.11: 68906207-68906207 on Build GRCh38
rs750773675 Chr.11: 68906129-68906129 on Build GRCh38
rs774522176 Chr.11: 68906167-68906167 on Build GRCh38
rs747952000 Chr.11: 68906337-68906337 on Build GRCh38
rs754319597 Chr.11: 68906197-68906197 on Build GRCh38
rs1057518943 Chr.11: 68906163-68906163 on Build GRCh38
rs117061430 Chr.11: 68906133-68906133 on Build GRCh38
rs183650590 Chr.11: 68906050-68906050 on Build GRCh38
rs863224880 Chr.11: 68906074-68906074 on Build GRCh38
rs375071662 Chr.11: 68906134-68906134 on Build GRCh38
rs926718557 Chr.11: 68906136-68906136 on Build GRCh38
rs768631087 Chr.11: 68906166-68906166 on Build GRCh38
rs1032941736 Chr.11: 68906053-68906053 on Build GRCh38
rs145237838 Chr.11: 68906217-68906217 on Build GRCh38
rs939987437 Chr.11: 68906358-68906358 on Build GRCh38
rs549801214 Chr.11: 68906233-68906233 on Build GRCh38
rs770746972 Chr.11: 68906151-68906151 on Build GRCh38
rs778774954 Chr.11: 68906211-68906211 on Build GRCh38
rs141376088 Chr.11: 68906312-68906312 on Build GRCh38
rs763529886 Chr.11: 68906304-68906304 on Build GRCh38
rs576739491 Chr.11: 68906047-68906047 on Build GRCh38
rs376644749 Chr.11: 68906199-68906199 on Build GRCh38
rs78807992 Chr.11: 68906114-68906114 on Build GRCh38
rs772261887 Chr.11: 68906259-68906259 on Build GRCh38
rs746660193 Chr.11: 68906216-68906216 on Build GRCh38
rs778432921 Chr.11: 68906141-68906141 on Build GRCh38
rs760896643 Chr.11: 68906268-68906268 on Build GRCh38

View Full Product Details