Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123593990-123594486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1B
Entrez Gene ID
57476
Gene Name
GRAM domain containing 1B
Gene Aliases
-
UniGene
Hs.144725
Gene Chromosome Location
Chr.11: 123358299-123627771 on Build GRCh38
Transcripts
NM_001286563.1 NM_001286564.1 NM_020716.2 XM_005271619.4 XM_005271621.4 XM_005271623.4 XM_005271624.4 XM_006718892.3 XM_011542926.2 XM_011542929.2 XM_011542930.2 XM_011542931.2 XM_011542933.1 XM_017018034.1 XM_017018035.1 XM_017018036.1 XM_017018037.1 XM_017018038.1 XM_017018039.1 XM_017018040.1 XM_017018041.1 XM_017018042.1 XM_017018043.1 XM_017018044.1 XM_017018045.1 XM_017018046.1 XM_017018047.1 XM_017018048.1 XM_017018049.1 XM_017018050.1 XM_017018051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
686770 c.285T>A p.N95K Chr.11: 123594111-123594111 on Build GRCh38
3808631 c.289G>T p.D97Y Chr.11: 123594115-123594115 on Build GRCh38
5749496 c.263G>A p.S88N Chr.11: 123594089-123594089 on Build GRCh38
1194582 c.299A>T p.K100M Chr.11: 123594125-123594125 on Build GRCh38
4018806 c.258G>A p.V86V Chr.11: 123594084-123594084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374199546 Chr.11: 123594180-123594180 on Build GRCh38
rs766015325 Chr.11: 123594153-123594153 on Build GRCh38
rs539067492 Chr.11: 123594255-123594255 on Build GRCh38
rs190410485 Chr.11: 123594052-123594052 on Build GRCh38
rs956913646 Chr.11: 123594193-123594193 on Build GRCh38
rs748574191 Chr.11: 123594045-123594045 on Build GRCh38
rs759608398 Chr.11: 123594090-123594090 on Build GRCh38
rs1035315848 Chr.11: 123594452-123594452 on Build GRCh38
rs763280370 Chr.11: 123594070-123594070 on Build GRCh38
rs765007012 Chr.11: 123594143-123594143 on Build GRCh38
rs928122144 Chr.11: 123594260-123594260 on Build GRCh38
rs368750346 Chr.11: 123594232-123594232 on Build GRCh38
rs766252761 Chr.11: 123594212-123594212 on Build GRCh38
rs751055464 Chr.11: 123594179-123594179 on Build GRCh38
rs778286471 Chr.11: 123594204-123594204 on Build GRCh38
rs377271879 Chr.11: 123594354-123594354 on Build GRCh38
rs781414768 Chr.11: 123594463-123594463 on Build GRCh38
rs373592130 Chr.11: 123594059-123594059 on Build GRCh38
rs74960781 Chr.11: 123594359-123594359 on Build GRCh38
rs905889649 Chr.11: 123594019-123594019 on Build GRCh38
rs781780945 Chr.11: 123594194-123594194 on Build GRCh38
rs558683060 Chr.11: 123594062-123594062 on Build GRCh38
rs1009379450 Chr.11: 123594156-123594156 on Build GRCh38
rs940440837 Chr.11: 123594318-123594318 on Build GRCh38
rs897392091 Chr.11: 123594048-123594048 on Build GRCh38
rs190982435 Chr.11: 123594240-123594240 on Build GRCh38
rs200747995 Chr.11: 123594293-123594293 on Build GRCh38
rs756630720 Chr.11: 123594201-123594201 on Build GRCh38
rs556730261 Chr.11: 123594292-123594292 on Build GRCh38
rs371977296 Chr.11: 123594219-123594219 on Build GRCh38
rs186909189 Chr.11: 123594216-123594216 on Build GRCh38
rs115344793 Chr.11: 123594328-123594328 on Build GRCh38
rs546227132 Chr.11: 123594444-123594444 on Build GRCh38
rs748651631 Chr.11: 123594198-123594198 on Build GRCh38
rs1022255388 Chr.11: 123594236-123594236 on Build GRCh38
rs2271631 Chr.11: 123594013-123594013 on Build GRCh38
rs905230649 Chr.11: 123594445-123594445 on Build GRCh38
rs1031125596 Chr.11: 123594184-123594184 on Build GRCh38
rs1052707115 Chr.11: 123594358-123594358 on Build GRCh38
rs538234160 Chr.11: 123594281-123594281 on Build GRCh38
rs758933182 Chr.11: 123594245-123594245 on Build GRCh38
rs553799720 Chr.11: 123594220-123594220 on Build GRCh38
rs1007024824 Chr.11: 123594377-123594377 on Build GRCh38
rs531490633 Chr.11: 123594149-123594149 on Build GRCh38
rs927345428 Chr.11: 123594316-123594316 on Build GRCh38
rs1022224306 Chr.11: 123594416-123594416 on Build GRCh38
rs907648453 Chr.11: 123594314-123594314 on Build GRCh38
rs1037349632 Chr.11: 123594323-123594323 on Build GRCh38
rs575773651 Chr.11: 123594284-123594284 on Build GRCh38
rs769076306 Chr.11: 123594033-123594033 on Build GRCh38
rs118067934 Chr.11: 123594078-123594078 on Build GRCh38
rs763984941 Chr.11: 123594094-123594094 on Build GRCh38
rs149364027 Chr.11: 123594044-123594044 on Build GRCh38
rs570959026 Chr.11: 123594110-123594110 on Build GRCh38
rs1002391950 Chr.11: 123594448-123594448 on Build GRCh38
rs961150147 Chr.11: 123594462-123594462 on Build GRCh38
rs890283926 Chr.11: 123594346-123594346 on Build GRCh38
rs557391590 Chr.11: 123594265-123594265 on Build GRCh38
rs920273788 Chr.11: 123594324-123594324 on Build GRCh38
rs776521709 Chr.11: 123594096-123594096 on Build GRCh38
rs182292408 Chr.11: 123594150-123594150 on Build GRCh38
rs1050359186 Chr.11: 123594331-123594331 on Build GRCh38
rs377023775 Chr.11: 123594077-123594077 on Build GRCh38
rs544398876 Chr.11: 123594286-123594286 on Build GRCh38
rs774530225 Chr.11: 123594302-123594302 on Build GRCh38
rs761665708 Chr.11: 123594108-123594108 on Build GRCh38
rs117594276 Chr.11: 123594399-123594399 on Build GRCh38

View Full Product Details