Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 133915203-133915704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF9B
Entrez Gene ID
22997
Gene Name
immunoglobulin superfamily member 9B
Gene Aliases
-
UniGene
Hs.204121
Gene Chromosome Location
Chr.11: 133908564-133956960 on Build GRCh38
Transcripts
NM_001277285.1 XM_006718795.3 XM_011542691.2 XM_011542692.2 XM_011542694.2 XM_011542695.2 XM_017017393.1 XM_017017395.1 XM_017017396.1

SNP Detail

SNP ID Chromosome Location
rs148582531 Chr.11: 133915268-133915268 on Build GRCh38
rs922285706 Chr.11: 133915645-133915645 on Build GRCh38
rs988901879 Chr.11: 133915562-133915562 on Build GRCh38
rs942937999 Chr.11: 133915639-133915639 on Build GRCh38
rs1023489999 Chr.11: 133915502-133915502 on Build GRCh38
rs502238 Chr.11: 133915268-133915268 on Build GRCh38
rs888168483 Chr.11: 133915676-133915676 on Build GRCh38
rs367567646 Chr.11: 133915578-133915578 on Build GRCh38
rs767011451 Chr.11: 133915257-133915260 on Build GRCh38
rs994206161 Chr.11: 133915669-133915669 on Build GRCh38
rs960136540 Chr.11: 133915325-133915325 on Build GRCh38
rs75766502 Chr.11: 133915289-133915289 on Build GRCh38
rs752318217 Chr.11: 133915257-133915258 on Build GRCh38
rs749074152 Chr.11: 133915623-133915623 on Build GRCh38
rs1000511863 Chr.11: 133915392-133915393 on Build GRCh38
rs1046729195 Chr.11: 133915660-133915660 on Build GRCh38
rs969296832 Chr.11: 133915252-133915252 on Build GRCh38
rs545837988 Chr.11: 133915238-133915238 on Build GRCh38
rs527672840 Chr.11: 133915678-133915678 on Build GRCh38
rs1008711829 Chr.11: 133915412-133915412 on Build GRCh38
rs1047286881 Chr.11: 133915475-133915475 on Build GRCh38
rs888929030 Chr.11: 133915398-133915398 on Build GRCh38
rs140270472 Chr.11: 133915369-133915369 on Build GRCh38
rs1008659391 Chr.11: 133915657-133915657 on Build GRCh38
rs369114905 Chr.11: 133915382-133915382 on Build GRCh38
rs1043211653 Chr.11: 133915367-133915367 on Build GRCh38
rs796969468 Chr.11: 133915268-133915269 on Build GRCh38
rs768443405 Chr.11: 133915656-133915656 on Build GRCh38
rs551368785 Chr.11: 133915527-133915527 on Build GRCh38
rs779909715 Chr.11: 133915621-133915621 on Build GRCh38
rs980074881 Chr.11: 133915253-133915253 on Build GRCh38
rs1041423004 Chr.11: 133915242-133915242 on Build GRCh38
rs913395234 Chr.11: 133915334-133915334 on Build GRCh38
rs758506742 Chr.11: 133915392-133915392 on Build GRCh38
rs994088756 Chr.11: 133915444-133915445 on Build GRCh38
rs900860116 Chr.11: 133915399-133915399 on Build GRCh38
rs1050758896 Chr.11: 133915644-133915644 on Build GRCh38
rs965450996 Chr.11: 133915434-133915435 on Build GRCh38
rs952778667 Chr.11: 133915476-133915476 on Build GRCh38
rs747844865 Chr.11: 133915267-133915268 on Build GRCh38
rs993929332 Chr.11: 133915474-133915474 on Build GRCh38
rs562650499 Chr.11: 133915491-133915491 on Build GRCh38
rs796664808 Chr.11: 133915354-133915354 on Build GRCh38
rs11223620 Chr.11: 133915455-133915455 on Build GRCh38
rs532230223 Chr.11: 133915270-133915270 on Build GRCh38
rs329677 Chr.11: 133915299-133915299 on Build GRCh38
rs1041320570 Chr.11: 133915640-133915640 on Build GRCh38
rs377026863 Chr.11: 133915268-133915269 on Build GRCh38
rs562222575 Chr.11: 133915413-133915413 on Build GRCh38
rs190940679 Chr.11: 133915504-133915504 on Build GRCh38
rs33964936 Chr.11: 133915269-133915270 on Build GRCh38
rs1023522699 Chr.11: 133915239-133915239 on Build GRCh38
rs904401315 Chr.11: 133915286-133915287 on Build GRCh38
rs1003488171 Chr.11: 133915517-133915517 on Build GRCh38
rs186869491 Chr.11: 133915490-133915490 on Build GRCh38
rs960353753 Chr.11: 133915528-133915528 on Build GRCh38
rs925606209 Chr.11: 133915572-133915572 on Build GRCh38
rs968850042 Chr.11: 133915608-133915608 on Build GRCh38
rs74438396 Chr.11: 133915266-133915266 on Build GRCh38
rs2086919 Chr.11: 133915443-133915443 on Build GRCh38
rs879149598 Chr.11: 133915638-133915638 on Build GRCh38
rs574210207 Chr.11: 133915480-133915480 on Build GRCh38
rs989322744 Chr.11: 133915330-133915330 on Build GRCh38
rs371697208 Chr.11: 133915679-133915679 on Build GRCh38
rs917031705 Chr.11: 133915232-133915232 on Build GRCh38
rs947509972 Chr.11: 133915353-133915353 on Build GRCh38
rs762753368 Chr.11: 133915259-133915260 on Build GRCh38
rs928631559 Chr.11: 133915255-133915255 on Build GRCh38
rs199957382 Chr.11: 133915266-133915268 on Build GRCh38
rs544362024 Chr.11: 133915374-133915374 on Build GRCh38
rs1040183772 Chr.11: 133915658-133915658 on Build GRCh38
rs1025767995 Chr.11: 133915673-133915673 on Build GRCh38

View Full Product Details