Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 133918866-133919359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF9B
Entrez Gene ID
22997
Gene Name
immunoglobulin superfamily member 9B
Gene Aliases
-
UniGene
Hs.204121
Gene Chromosome Location
Chr.11: 133908564-133956960 on Build GRCh38
Transcripts
NM_001277285.1 XM_006718795.3 XM_011542691.2 XM_011542692.2 XM_011542694.2 XM_011542695.2 XM_017017393.1 XM_017017394.1 XM_017017395.1 XM_017017396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1352696 c.2380G>A p.A794T Chr.11: 133919077-133919077 on Build GRCh38
4806484 c.2366A>T p.D789V Chr.11: 133919091-133919091 on Build GRCh38
1604322 c.2377C>G p.P793A Chr.11: 133919080-133919080 on Build GRCh38
925074 c.2352G>A p.G784G Chr.11: 133919105-133919105 on Build GRCh38
925072 c.2363G>A p.R788K Chr.11: 133919094-133919094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543146110 Chr.11: 133919279-133919279 on Build GRCh38
rs976083666 Chr.11: 133919198-133919198 on Build GRCh38
rs575747028 Chr.11: 133919146-133919146 on Build GRCh38
rs749297626 Chr.11: 133919094-133919094 on Build GRCh38
rs547713732 Chr.11: 133918960-133918960 on Build GRCh38
rs766389662 Chr.11: 133919099-133919099 on Build GRCh38
rs995403395 Chr.11: 133918931-133918931 on Build GRCh38
rs774179139 Chr.11: 133919109-133919109 on Build GRCh38
rs868046085 Chr.11: 133919333-133919333 on Build GRCh38
rs1035923804 Chr.11: 133919122-133919122 on Build GRCh38
rs566347098 Chr.11: 133918961-133918961 on Build GRCh38
rs12801289 Chr.11: 133919304-133919304 on Build GRCh38
rs771394116 Chr.11: 133919119-133919120 on Build GRCh38
rs751412549 Chr.11: 133919020-133919020 on Build GRCh38
rs377573270 Chr.11: 133919005-133919005 on Build GRCh38
rs564581302 Chr.11: 133918914-133918914 on Build GRCh38
rs918358961 Chr.11: 133919301-133919301 on Build GRCh38
rs1026929272 Chr.11: 133918936-133918936 on Build GRCh38
rs932580977 Chr.11: 133918959-133918959 on Build GRCh38
rs373119596 Chr.11: 133919001-133919001 on Build GRCh38
rs770380757 Chr.11: 133919135-133919135 on Build GRCh38
rs773228025 Chr.11: 133919014-133919014 on Build GRCh38
rs200483863 Chr.11: 133919129-133919129 on Build GRCh38
rs981639042 Chr.11: 133918902-133918902 on Build GRCh38
rs779285995 Chr.11: 133918917-133918918 on Build GRCh38
rs747819474 Chr.11: 133919086-133919086 on Build GRCh38
rs750342470 Chr.11: 133918994-133918999 on Build GRCh38
rs1052730627 Chr.11: 133919295-133919295 on Build GRCh38
rs765176010 Chr.11: 133919122-133919123 on Build GRCh38
rs189331775 Chr.11: 133918992-133918992 on Build GRCh38
rs1049240570 Chr.11: 133919218-133919218 on Build GRCh38
rs1002927642 Chr.11: 133919292-133919292 on Build GRCh38
rs771494094 Chr.11: 133919258-133919259 on Build GRCh38
rs888016995 Chr.11: 133919052-133919052 on Build GRCh38
rs572279411 Chr.11: 133919158-133919158 on Build GRCh38
rs756267848 Chr.11: 133919024-133919024 on Build GRCh38
rs762080294 Chr.11: 133919120-133919120 on Build GRCh38
rs774280318 Chr.11: 133919136-133919136 on Build GRCh38
rs759338265 Chr.11: 133919112-133919112 on Build GRCh38
rs751674756 Chr.11: 133919126-133919127 on Build GRCh38
rs755315214 Chr.11: 133919124-133919124 on Build GRCh38
rs549377071 Chr.11: 133918908-133918908 on Build GRCh38
rs1037158038 Chr.11: 133919296-133919296 on Build GRCh38
rs896856712 Chr.11: 133918919-133918919 on Build GRCh38
rs200886253 Chr.11: 133919077-133919077 on Build GRCh38
rs780225131 Chr.11: 133919048-133919049 on Build GRCh38
rs754000114 Chr.11: 133919119-133919119 on Build GRCh38
rs914465168 Chr.11: 133918928-133918928 on Build GRCh38
rs561099249 Chr.11: 133919234-133919234 on Build GRCh38
rs12800905 Chr.11: 133919097-133919097 on Build GRCh38
rs1002551890 Chr.11: 133919080-133919080 on Build GRCh38
rs769613312 Chr.11: 133919126-133919126 on Build GRCh38
rs554121697 Chr.11: 133919137-133919137 on Build GRCh38
rs761354502 Chr.11: 133919007-133919007 on Build GRCh38
rs373112099 Chr.11: 133919090-133919090 on Build GRCh38
rs985712174 Chr.11: 133919202-133919202 on Build GRCh38
rs780558285 Chr.11: 133919123-133919123 on Build GRCh38
rs763046193 Chr.11: 133919016-133919016 on Build GRCh38
rs756305133 Chr.11: 133919129-133919130 on Build GRCh38
rs1007855863 Chr.11: 133918909-133918910 on Build GRCh38
rs948992275 Chr.11: 133919264-133919264 on Build GRCh38
rs751591483 Chr.11: 133919089-133919090 on Build GRCh38
rs537591777 Chr.11: 133919075-133919075 on Build GRCh38
rs913890505 Chr.11: 133918986-133918988 on Build GRCh38
rs925501394 Chr.11: 133919177-133919177 on Build GRCh38
rs548717168 Chr.11: 133918986-133918986 on Build GRCh38
rs10791338 Chr.11: 133918974-133918974 on Build GRCh38
rs912280462 Chr.11: 133919053-133919053 on Build GRCh38
rs905560409 Chr.11: 133919329-133919329 on Build GRCh38
rs778276561 Chr.11: 133919119-133919124 on Build GRCh38
rs747446654 Chr.11: 133919082-133919082 on Build GRCh38
rs559772849 Chr.11: 133919119-133919119 on Build GRCh38
rs776380242 Chr.11: 133919085-133919085 on Build GRCh38
rs754091899 Chr.11: 133919069-133919069 on Build GRCh38
rs979420892 Chr.11: 133919172-133919172 on Build GRCh38
rs919314205 Chr.11: 133919203-133919203 on Build GRCh38
rs765295237 Chr.11: 133919121-133919121 on Build GRCh38
rs59914981 Chr.11: 133919128-133919129 on Build GRCh38
rs969847603 Chr.11: 133918953-133918953 on Build GRCh38
rs867099937 Chr.11: 133919302-133919302 on Build GRCh38
rs752868696 Chr.11: 133919127-133919128 on Build GRCh38
rs764846987 Chr.11: 133919008-133919008 on Build GRCh38
rs971153252 Chr.11: 133918901-133918901 on Build GRCh38
rs989942539 Chr.11: 133918912-133918912 on Build GRCh38
rs761728360 Chr.11: 133919121-133919122 on Build GRCh38
rs746633877 Chr.11: 133918995-133918995 on Build GRCh38
rs910544975 Chr.11: 133919145-133919145 on Build GRCh38
rs771839413 Chr.11: 133919154-133919154 on Build GRCh38
rs577336826 Chr.11: 133919083-133919083 on Build GRCh38
rs780247015 Chr.11: 133919078-133919078 on Build GRCh38
rs992289913 Chr.11: 133918980-133918980 on Build GRCh38
rs543160427 Chr.11: 133919337-133919337 on Build GRCh38
rs1045010875 Chr.11: 133919282-133919282 on Build GRCh38
rs775132410 Chr.11: 133919120-133919121 on Build GRCh38
rs557402535 Chr.11: 133919124-133919125 on Build GRCh38
rs781729309 Chr.11: 133919125-133919125 on Build GRCh38
rs531979452 Chr.11: 133918918-133918918 on Build GRCh38
rs764330267 Chr.11: 133919029-133919029 on Build GRCh38
rs775980946 Chr.11: 133918993-133918993 on Build GRCh38
rs767306266 Chr.11: 133919117-133919117 on Build GRCh38
rs766086261 Chr.11: 133919017-133919017 on Build GRCh38
rs909420369 Chr.11: 133919243-133919243 on Build GRCh38
rs542464048 Chr.11: 133919199-133919199 on Build GRCh38
rs929405873 Chr.11: 133919212-133919212 on Build GRCh38
rs746361407 Chr.11: 133919076-133919076 on Build GRCh38
rs907456836 Chr.11: 133919288-133919288 on Build GRCh38
rs1033724405 Chr.11: 133918966-133918966 on Build GRCh38
rs374455300 Chr.11: 133918907-133918907 on Build GRCh38
rs1021410613 Chr.11: 133919322-133919322 on Build GRCh38
rs565660801 Chr.11: 133919038-133919038 on Build GRCh38
rs758390652 Chr.11: 133919041-133919042 on Build GRCh38
rs757623581 Chr.11: 133919118-133919119 on Build GRCh38
rs766622115 Chr.11: 133919072-133919072 on Build GRCh38
rs954304461 Chr.11: 133919191-133919191 on Build GRCh38
rs1013609390 Chr.11: 133919065-133919065 on Build GRCh38

View Full Product Details