Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2820008-2820516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITFG2
Entrez Gene ID
55846
Gene Name
integrin alpha FG-GAP repeat containing 2
Gene Aliases
MDS028
UniGene
Hs.446098
Gene Chromosome Location
Chr.12: 2812621-2832217 on Build GRCh38
Transcripts
NM_018463.3 NR_130744.1 XM_005253707.4 XM_005253709.4 XM_017019641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4497204 c.155C>T p.P52L Chr.12: 2820170-2820170 on Build GRCh38
5226199 c.165G>A p.L55L Chr.12: 2820180-2820180 on Build GRCh38
2204692 c.463T>G p.F155V Chr.12: 2820142-2820142 on Build GRCh38
4041293 c.188A>C p.K63T Chr.12: 2820203-2820203 on Build GRCh38
6306616 c.99G>T p.V33V Chr.12: 2820114-2820114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019391908 Chr.12: 2820367-2820367 on Build GRCh38
rs925186860 Chr.12: 2820340-2820340 on Build GRCh38
rs373513457 Chr.12: 2820038-2820038 on Build GRCh38
rs564138795 Chr.12: 2820388-2820388 on Build GRCh38
rs772137151 Chr.12: 2820150-2820150 on Build GRCh38
rs781024331 Chr.12: 2820272-2820272 on Build GRCh38
rs778907558 Chr.12: 2820134-2820134 on Build GRCh38
rs766772907 Chr.12: 2820070-2820070 on Build GRCh38
rs374022306 Chr.12: 2820036-2820036 on Build GRCh38
rs1033828334 Chr.12: 2820451-2820451 on Build GRCh38
rs530516097 Chr.12: 2820384-2820384 on Build GRCh38
rs919556987 Chr.12: 2820114-2820114 on Build GRCh38
rs370623891 Chr.12: 2820076-2820076 on Build GRCh38
rs919432381 Chr.12: 2820433-2820433 on Build GRCh38
rs781245944 Chr.12: 2820089-2820089 on Build GRCh38
rs563385979 Chr.12: 2820079-2820079 on Build GRCh38
rs760174850 Chr.12: 2820040-2820040 on Build GRCh38
rs760259556 Chr.12: 2820159-2820159 on Build GRCh38
rs993786898 Chr.12: 2820204-2820204 on Build GRCh38
rs762825809 Chr.12: 2820467-2820467 on Build GRCh38
rs984928558 Chr.12: 2820239-2820239 on Build GRCh38
rs554016203 Chr.12: 2820146-2820146 on Build GRCh38
rs1026967952 Chr.12: 2820210-2820210 on Build GRCh38
rs1056478834 Chr.12: 2820062-2820062 on Build GRCh38
rs778087371 Chr.12: 2820104-2820104 on Build GRCh38
rs944919907 Chr.12: 2820065-2820065 on Build GRCh38
rs191657317 Chr.12: 2820420-2820420 on Build GRCh38
rs376269449 Chr.12: 2820145-2820145 on Build GRCh38
rs770825251 Chr.12: 2820129-2820129 on Build GRCh38
rs761504082 Chr.12: 2820182-2820182 on Build GRCh38
rs892938916 Chr.12: 2820334-2820334 on Build GRCh38
rs186352072 Chr.12: 2820355-2820355 on Build GRCh38
rs566752715 Chr.12: 2820453-2820453 on Build GRCh38
rs377000219 Chr.12: 2820046-2820046 on Build GRCh38
rs998740817 Chr.12: 2820281-2820281 on Build GRCh38
rs755253553 Chr.12: 2820436-2820436 on Build GRCh38
rs759969136 Chr.12: 2820213-2820213 on Build GRCh38
rs756185712 Chr.12: 2820235-2820235 on Build GRCh38
rs373485572 Chr.12: 2820048-2820048 on Build GRCh38
rs889542232 Chr.12: 2820138-2820138 on Build GRCh38
rs1049596681 Chr.12: 2820346-2820346 on Build GRCh38
rs752677731 Chr.12: 2820221-2820221 on Build GRCh38
rs985065539 Chr.12: 2820466-2820466 on Build GRCh38
rs755538213 Chr.12: 2820085-2820085 on Build GRCh38
rs959464103 Chr.12: 2820262-2820262 on Build GRCh38
rs761929643 Chr.12: 2820376-2820376 on Build GRCh38
rs1041171702 Chr.12: 2820357-2820357 on Build GRCh38
rs756360579 Chr.12: 2820262-2820266 on Build GRCh38
rs544245312 Chr.12: 2820387-2820387 on Build GRCh38
rs71458018 Chr.12: 2820247-2820247 on Build GRCh38
rs369652729 Chr.12: 2820047-2820047 on Build GRCh38
rs888039011 Chr.12: 2820385-2820385 on Build GRCh38
rs764126565 Chr.12: 2820238-2820238 on Build GRCh38
rs751988994 Chr.12: 2820197-2820197 on Build GRCh38
rs768156895 Chr.12: 2820175-2820175 on Build GRCh38
rs766717797 Chr.12: 2820195-2820195 on Build GRCh38
rs896845521 Chr.12: 2820361-2820361 on Build GRCh38
rs752980685 Chr.12: 2820079-2820079 on Build GRCh38
rs529342178 Chr.12: 2820464-2820464 on Build GRCh38
rs764587259 Chr.12: 2820276-2820276 on Build GRCh38
rs749511053 Chr.12: 2820107-2820107 on Build GRCh38
rs761880536 Chr.12: 2820334-2820335 on Build GRCh38
rs370019130 Chr.12: 2820035-2820035 on Build GRCh38
rs768090911 Chr.12: 2820220-2820220 on Build GRCh38
rs948783103 Chr.12: 2820487-2820487 on Build GRCh38
rs952255073 Chr.12: 2820230-2820230 on Build GRCh38
rs774801587 Chr.12: 2820156-2820156 on Build GRCh38
rs907273761 Chr.12: 2820280-2820280 on Build GRCh38
rs775128534 Chr.12: 2820039-2820039 on Build GRCh38
rs150963406 Chr.12: 2820417-2820417 on Build GRCh38
rs754075878 Chr.12: 2820253-2820253 on Build GRCh38
rs776318594 Chr.12: 2820178-2820178 on Build GRCh38
rs1031927738 Chr.12: 2820308-2820308 on Build GRCh38
rs903717969 Chr.12: 2820100-2820100 on Build GRCh38
rs894493117 Chr.12: 2820243-2820243 on Build GRCh38
rs1011462537 Chr.12: 2820341-2820341 on Build GRCh38
rs572677865 Chr.12: 2820055-2820055 on Build GRCh38
rs181333125 Chr.12: 2820351-2820351 on Build GRCh38
rs972470978 Chr.12: 2820422-2820422 on Build GRCh38
rs916152887 Chr.12: 2820477-2820477 on Build GRCh38
rs986856201 Chr.12: 2820362-2820362 on Build GRCh38
rs373063840 Chr.12: 2820101-2820101 on Build GRCh38
rs751478384 Chr.12: 2820141-2820141 on Build GRCh38
rs959514389 Chr.12: 2820452-2820452 on Build GRCh38
rs757047982 Chr.12: 2820252-2820252 on Build GRCh38
rs372802471 Chr.12: 2820042-2820042 on Build GRCh38
rs189870336 Chr.12: 2820257-2820257 on Build GRCh38
rs371263886 Chr.12: 2820254-2820254 on Build GRCh38
rs917881724 Chr.12: 2820066-2820066 on Build GRCh38
rs185016186 Chr.12: 2820219-2820219 on Build GRCh38
rs961308287 Chr.12: 2820188-2820188 on Build GRCh38
rs978499456 Chr.12: 2820478-2820478 on Build GRCh38
rs945357390 Chr.12: 2820299-2820299 on Build GRCh38

View Full Product Details