Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2820584-2821090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITFG2
Entrez Gene ID
55846
Gene Name
integrin alpha FG-GAP repeat containing 2
Gene Aliases
MDS028
UniGene
Hs.446098
Gene Chromosome Location
Chr.12: 2812621-2832217 on Build GRCh38
Transcripts
NM_018463.3 NR_130744.1 XM_005253707.4 XM_005253709.4 XM_017019641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4041294 c.618G>A p.A206A Chr.12: 2820795-2820795 on Build GRCh38
4041296 c.666T>A p.P222P Chr.12: 2820843-2820843 on Build GRCh38
5758766 c.681C>T p.P227P Chr.12: 2820858-2820858 on Build GRCh38
7314674 c.331G>T p.A111S Chr.12: 2820844-2820844 on Build GRCh38
6335829 c.581G>T p.G194V Chr.12: 2820758-2820758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019700541 Chr.12: 2820660-2820660 on Build GRCh38
rs752670394 Chr.12: 2820909-2820909 on Build GRCh38
rs981684476 Chr.12: 2820810-2820810 on Build GRCh38
rs750207163 Chr.12: 2820815-2820815 on Build GRCh38
rs144639569 Chr.12: 2820773-2820773 on Build GRCh38
rs1026770295 Chr.12: 2821036-2821036 on Build GRCh38
rs1032414915 Chr.12: 2821015-2821015 on Build GRCh38
rs148451445 Chr.12: 2820802-2820802 on Build GRCh38
rs1009859142 Chr.12: 2820651-2820651 on Build GRCh38
rs71458020 Chr.12: 2820976-2820976 on Build GRCh38
rs772237742 Chr.12: 2820868-2820868 on Build GRCh38
rs540414717 Chr.12: 2820898-2820898 on Build GRCh38
rs562755723 Chr.12: 2821040-2821040 on Build GRCh38
rs1033880817 Chr.12: 2820655-2820655 on Build GRCh38
rs543991183 Chr.12: 2820841-2820841 on Build GRCh38
rs889576016 Chr.12: 2820925-2820925 on Build GRCh38
rs772468159 Chr.12: 2820855-2820855 on Build GRCh38
rs780687011 Chr.12: 2820823-2820823 on Build GRCh38
rs577942766 Chr.12: 2820897-2820897 on Build GRCh38
rs545766779 Chr.12: 2820813-2820813 on Build GRCh38
rs977000036 Chr.12: 2820826-2820826 on Build GRCh38
rs999634157 Chr.12: 2821006-2821006 on Build GRCh38
rs934658128 Chr.12: 2820822-2820822 on Build GRCh38
rs776042281 Chr.12: 2820792-2820792 on Build GRCh38
rs368269395 Chr.12: 2820700-2820700 on Build GRCh38
rs759924227 Chr.12: 2820771-2820771 on Build GRCh38
rs575134327 Chr.12: 2820821-2820821 on Build GRCh38
rs764518740 Chr.12: 2820866-2820866 on Build GRCh38
rs778780708 Chr.12: 2820686-2820686 on Build GRCh38
rs896831616 Chr.12: 2820961-2820961 on Build GRCh38
rs529172554 Chr.12: 2820954-2820954 on Build GRCh38
rs140728858 Chr.12: 2820752-2820752 on Build GRCh38
rs370764849 Chr.12: 2820793-2820793 on Build GRCh38
rs199914287 Chr.12: 2820693-2820693 on Build GRCh38
rs769396343 Chr.12: 2820840-2820840 on Build GRCh38
rs373908169 Chr.12: 2820795-2820795 on Build GRCh38
rs755599196 Chr.12: 2820921-2820921 on Build GRCh38
rs71458019 Chr.12: 2820913-2820913 on Build GRCh38
rs1053979166 Chr.12: 2820644-2820644 on Build GRCh38
rs560476024 Chr.12: 2820928-2820928 on Build GRCh38
rs551817666 Chr.12: 2820633-2820633 on Build GRCh38
rs142638240 Chr.12: 2820816-2820816 on Build GRCh38
rs775781549 Chr.12: 2820860-2820860 on Build GRCh38
rs776065268 Chr.12: 2820703-2820703 on Build GRCh38
rs573983315 Chr.12: 2820746-2820746 on Build GRCh38
rs750654831 Chr.12: 2820880-2820880 on Build GRCh38
rs926404295 Chr.12: 2820634-2820634 on Build GRCh38
rs765451678 Chr.12: 2820879-2820879 on Build GRCh38
rs200954465 Chr.12: 2820744-2820744 on Build GRCh38
rs952439187 Chr.12: 2821048-2821048 on Build GRCh38
rs202152027 Chr.12: 2820694-2820694 on Build GRCh38
rs150956987 Chr.12: 2820842-2820842 on Build GRCh38
rs929709371 Chr.12: 2820629-2820629 on Build GRCh38
rs948991029 Chr.12: 2820799-2820799 on Build GRCh38
rs549213209 Chr.12: 2821031-2821031 on Build GRCh38
rs893978405 Chr.12: 2820648-2820648 on Build GRCh38
rs747671837 Chr.12: 2820825-2820825 on Build GRCh38
rs757493974 Chr.12: 2820805-2820805 on Build GRCh38
rs902655098 Chr.12: 2821005-2821005 on Build GRCh38
rs368011279 Chr.12: 2820882-2820882 on Build GRCh38
rs2968908 Chr.12: 2820861-2820861 on Build GRCh38
rs781130955 Chr.12: 2820908-2820908 on Build GRCh38
rs937742277 Chr.12: 2820640-2820640 on Build GRCh38
rs1048064362 Chr.12: 2820632-2820632 on Build GRCh38
rs140856468 Chr.12: 2820846-2820846 on Build GRCh38
rs1048116298 Chr.12: 2820858-2820858 on Build GRCh38
rs1047012316 Chr.12: 2821000-2821000 on Build GRCh38
rs1006452738 Chr.12: 2820639-2820639 on Build GRCh38
rs566892011 Chr.12: 2820849-2820849 on Build GRCh38
rs761007706 Chr.12: 2820735-2820735 on Build GRCh38
rs201767066 Chr.12: 2820697-2820697 on Build GRCh38
rs766193546 Chr.12: 2820891-2820891 on Build GRCh38

View Full Product Details