Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14777035-14777578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf60
Entrez Gene ID
144608
Gene Name
chromosome 12 open reading frame 60
Gene Aliases
-
UniGene
Hs.659951
Gene Chromosome Location
Chr.12: 14567991-14823858 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_177925.3 NR_027716.2

Gene Symbol
H2AFJ
Entrez Gene ID
55766
Gene Name
H2A histone family member J
Gene Aliases
H2AJ
UniGene
Hs.524280
Gene Chromosome Location
Chr.12: 14774336-14778002 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_177925.3 NR_027716.2

SNP Detail

SNP ID Chromosome Location
rs911990900 Chr.12: 14777406-14777406 on Build GRCh38
rs1015152028 Chr.12: 14777257-14777257 on Build GRCh38
rs973583883 Chr.12: 14777327-14777327 on Build GRCh38
rs138850700 Chr.12: 14777095-14777095 on Build GRCh38
rs999350599 Chr.12: 14777095-14777095 on Build GRCh38
rs7967703 Chr.12: 14777529-14777529 on Build GRCh38
rs982197165 Chr.12: 14777215-14777215 on Build GRCh38
rs560314969 Chr.12: 14777229-14777229 on Build GRCh38
rs1019963385 Chr.12: 14777073-14777074 on Build GRCh38
rs1039959781 Chr.12: 14777301-14777301 on Build GRCh38
rs397841360 Chr.12: 14777104-14777104 on Build GRCh38
rs564404190 Chr.12: 14777535-14777535 on Build GRCh38
rs888266384 Chr.12: 14777249-14777249 on Build GRCh38
rs970346635 Chr.12: 14777190-14777190 on Build GRCh38
rs918621826 Chr.12: 14777149-14777149 on Build GRCh38
rs7980962 Chr.12: 14777178-14777178 on Build GRCh38
rs962631470 Chr.12: 14777309-14777309 on Build GRCh38
rs765616344 Chr.12: 14777164-14777164 on Build GRCh38
rs865984064 Chr.12: 14777313-14777313 on Build GRCh38
rs902448485 Chr.12: 14777090-14777090 on Build GRCh38
rs186800409 Chr.12: 14777130-14777130 on Build GRCh38
rs986145354 Chr.12: 14777391-14777391 on Build GRCh38
rs7952904 Chr.12: 14777356-14777356 on Build GRCh38
rs34712595 Chr.12: 14777334-14777335 on Build GRCh38
rs920802097 Chr.12: 14777335-14777335 on Build GRCh38
rs192265844 Chr.12: 14777541-14777541 on Build GRCh38
rs990964586 Chr.12: 14777506-14777506 on Build GRCh38
rs902889193 Chr.12: 14777351-14777351 on Build GRCh38
rs1053622026 Chr.12: 14777504-14777504 on Build GRCh38
rs750747609 Chr.12: 14777375-14777375 on Build GRCh38
rs978540770 Chr.12: 14777078-14777078 on Build GRCh38
rs777048212 Chr.12: 14777089-14777089 on Build GRCh38
rs906299361 Chr.12: 14777544-14777544 on Build GRCh38
rs757492743 Chr.12: 14777062-14777062 on Build GRCh38
rs568929073 Chr.12: 14777555-14777555 on Build GRCh38
rs558639022 Chr.12: 14777510-14777510 on Build GRCh38
rs1048499227 Chr.12: 14777224-14777224 on Build GRCh38
rs562475753 Chr.12: 14777120-14777120 on Build GRCh38
rs932155079 Chr.12: 14777350-14777350 on Build GRCh38
rs577511793 Chr.12: 14777067-14777067 on Build GRCh38
rs3838353 Chr.12: 14777104-14777105 on Build GRCh38
rs7967694 Chr.12: 14777503-14777503 on Build GRCh38
rs944702404 Chr.12: 14777453-14777453 on Build GRCh38
rs999928148 Chr.12: 14777355-14777355 on Build GRCh38
rs760645530 Chr.12: 14777061-14777062 on Build GRCh38
rs982045185 Chr.12: 14777142-14777142 on Build GRCh38
rs148244039 Chr.12: 14777389-14777390 on Build GRCh38
rs1024232895 Chr.12: 14777507-14777507 on Build GRCh38
rs144484024 Chr.12: 14777387-14777388 on Build GRCh38
rs7967360 Chr.12: 14777275-14777275 on Build GRCh38
rs796593618 Chr.12: 14777333-14777334 on Build GRCh38
rs1019953553 Chr.12: 14777344-14777344 on Build GRCh38
rs1032620943 Chr.12: 14777110-14777110 on Build GRCh38
rs923869818 Chr.12: 14777478-14777478 on Build GRCh38
rs3789999 Chr.12: 14777121-14777121 on Build GRCh38
rs958719581 Chr.12: 14777500-14777500 on Build GRCh38
rs1042136210 Chr.12: 14777462-14777462 on Build GRCh38
rs567160588 Chr.12: 14777183-14777184 on Build GRCh38
rs550913807 Chr.12: 14777543-14777543 on Build GRCh38
rs544146654 Chr.12: 14777108-14777108 on Build GRCh38
rs111846873 Chr.12: 14777553-14777553 on Build GRCh38
rs199736949 Chr.12: 14777349-14777350 on Build GRCh38
rs7964370 Chr.12: 14777063-14777063 on Build GRCh38
rs1040940637 Chr.12: 14777076-14777076 on Build GRCh38
rs944718173 Chr.12: 14777072-14777072 on Build GRCh38
rs527464864 Chr.12: 14777247-14777247 on Build GRCh38
rs117925560 Chr.12: 14777481-14777481 on Build GRCh38

View Full Product Details