Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6040426-6040922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB2
Entrez Gene ID
8514
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 2
Gene Aliases
AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, KCNA2B, KV-BETA-2
UniGene
Hs.440497
Gene Chromosome Location
Chr.1: 5992298-6101193 on Build GRCh38
Transcripts
NM_001199860.1 NM_001199861.1 NM_003636.3 NM_172130.2 XM_005263514.2 XM_011542321.2 XM_011542322.2 XM_017002618.1 XM_017002619.1 XM_017002621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4009153 c.53C>T p.T18M Chr.1: 6040621-6040621 on Build GRCh38
6690512 c.47G>A p.R16Q Chr.1: 6040615-6040615 on Build GRCh38
6598070 c.21G>A p.T7T Chr.1: 6040589-6040589 on Build GRCh38
6056075 c.56G>A p.G19D Chr.1: 6040624-6040624 on Build GRCh38
6598069 c.21G>A p.T7T Chr.1: 6040589-6040589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749935103 Chr.1: 6040607-6040607 on Build GRCh38
rs552829930 Chr.1: 6040663-6040663 on Build GRCh38
rs920921615 Chr.1: 6040866-6040866 on Build GRCh38
rs773699435 Chr.1: 6040674-6040674 on Build GRCh38
rs749860065 Chr.1: 6040735-6040735 on Build GRCh38
rs370596239 Chr.1: 6040642-6040642 on Build GRCh38
rs371095544 Chr.1: 6040563-6040563 on Build GRCh38
rs557476385 Chr.1: 6040738-6040738 on Build GRCh38
rs1011661565 Chr.1: 6040800-6040800 on Build GRCh38
rs4908494 Chr.1: 6040795-6040795 on Build GRCh38
rs1048604276 Chr.1: 6040542-6040542 on Build GRCh38
rs781493823 Chr.1: 6040602-6040602 on Build GRCh38
rs777718700 Chr.1: 6040632-6040632 on Build GRCh38
rs377567401 Chr.1: 6040557-6040557 on Build GRCh38
rs1036955302 Chr.1: 6040771-6040771 on Build GRCh38
rs912443455 Chr.1: 6040720-6040720 on Build GRCh38
rs201737295 Chr.1: 6040622-6040622 on Build GRCh38
rs541930222 Chr.1: 6040589-6040589 on Build GRCh38
rs563820808 Chr.1: 6040598-6040598 on Build GRCh38
rs571253296 Chr.1: 6040850-6040850 on Build GRCh38
rs937424131 Chr.1: 6040460-6040460 on Build GRCh38
rs759796433 Chr.1: 6040751-6040752 on Build GRCh38
rs138650831 Chr.1: 6040586-6040586 on Build GRCh38
rs1037197744 Chr.1: 6040776-6040776 on Build GRCh38
rs924648964 Chr.1: 6040505-6040505 on Build GRCh38
rs1005010560 Chr.1: 6040770-6040770 on Build GRCh38
rs762222888 Chr.1: 6040679-6040684 on Build GRCh38
rs368235462 Chr.1: 6040669-6040669 on Build GRCh38
rs968656011 Chr.1: 6040851-6040851 on Build GRCh38
rs748155676 Chr.1: 6040623-6040623 on Build GRCh38
rs947173532 Chr.1: 6040484-6040484 on Build GRCh38
rs2294937 Chr.1: 6040665-6040665 on Build GRCh38
rs755694345 Chr.1: 6040529-6040529 on Build GRCh38
rs754826043 Chr.1: 6040621-6040621 on Build GRCh38
rs967870581 Chr.1: 6040807-6040807 on Build GRCh38
rs921325197 Chr.1: 6040882-6040882 on Build GRCh38
rs1034334888 Chr.1: 6040794-6040794 on Build GRCh38
rs547329290 Chr.1: 6040748-6040748 on Build GRCh38
rs748273253 Chr.1: 6040631-6040631 on Build GRCh38
rs1003563071 Chr.1: 6040778-6040778 on Build GRCh38
rs201381401 Chr.1: 6040539-6040539 on Build GRCh38
rs977508892 Chr.1: 6040495-6040495 on Build GRCh38
rs376203067 Chr.1: 6040614-6040614 on Build GRCh38
rs762200918 Chr.1: 6040666-6040666 on Build GRCh38
rs775798232 Chr.1: 6040596-6040596 on Build GRCh38
rs35719658 Chr.1: 6040609-6040610 on Build GRCh38
rs758296656 Chr.1: 6040624-6040624 on Build GRCh38
rs1056851624 Chr.1: 6040789-6040789 on Build GRCh38
rs375390362 Chr.1: 6040610-6040610 on Build GRCh38
rs766108683 Chr.1: 6040656-6040669 on Build GRCh38
rs753537552 Chr.1: 6040697-6040697 on Build GRCh38
rs576762094 Chr.1: 6040759-6040759 on Build GRCh38
rs200939954 Chr.1: 6040585-6040585 on Build GRCh38
rs777687172 Chr.1: 6040728-6040728 on Build GRCh38
rs954188914 Chr.1: 6040891-6040891 on Build GRCh38
rs760166516 Chr.1: 6040692-6040692 on Build GRCh38
rs369939805 Chr.1: 6040702-6040702 on Build GRCh38
rs1031945761 Chr.1: 6040861-6040861 on Build GRCh38
rs541024524 Chr.1: 6040478-6040478 on Build GRCh38
rs758297152 Chr.1: 6040603-6040603 on Build GRCh38
rs767097088 Chr.1: 6040604-6040604 on Build GRCh38
rs1048636315 Chr.1: 6040752-6040752 on Build GRCh38
rs145359975 Chr.1: 6040715-6040715 on Build GRCh38
rs965217666 Chr.1: 6040870-6040870 on Build GRCh38
rs912398306 Chr.1: 6040835-6040835 on Build GRCh38
rs912207209 Chr.1: 6040541-6040541 on Build GRCh38
rs981856350 Chr.1: 6040449-6040449 on Build GRCh38
rs751338529 Chr.1: 6040467-6040467 on Build GRCh38
rs943685277 Chr.1: 6040543-6040543 on Build GRCh38
rs921282477 Chr.1: 6040716-6040716 on Build GRCh38
rs984126579 Chr.1: 6040894-6040894 on Build GRCh38
rs774613467 Chr.1: 6040669-6040669 on Build GRCh38
rs943972757 Chr.1: 6040722-6040722 on Build GRCh38
rs914438704 Chr.1: 6040511-6040511 on Build GRCh38
rs977260041 Chr.1: 6040816-6040816 on Build GRCh38
rs750142415 Chr.1: 6040528-6040528 on Build GRCh38
rs200252793 Chr.1: 6040530-6040531 on Build GRCh38
rs775922659 Chr.1: 6040648-6040648 on Build GRCh38
rs780557707 Chr.1: 6040523-6040523 on Build GRCh38
rs958762441 Chr.1: 6040678-6040678 on Build GRCh38
rs975340580 Chr.1: 6040854-6040854 on Build GRCh38
rs767921441 Chr.1: 6040671-6040671 on Build GRCh38
rs977668551 Chr.1: 6040719-6040719 on Build GRCh38
rs763416022 Chr.1: 6040597-6040597 on Build GRCh38
rs528832984 Chr.1: 6040703-6040703 on Build GRCh38
rs779683151 Chr.1: 6040613-6040613 on Build GRCh38
rs113892791 Chr.1: 6040696-6040696 on Build GRCh38
rs749091483 Chr.1: 6040538-6040538 on Build GRCh38
rs944132453 Chr.1: 6040527-6040527 on Build GRCh38
rs547946318 Chr.1: 6040876-6040876 on Build GRCh38
rs749513241 Chr.1: 6040654-6040654 on Build GRCh38
rs888628906 Chr.1: 6040547-6040547 on Build GRCh38
rs377145481 Chr.1: 6040533-6040533 on Build GRCh38
rs559281538 Chr.1: 6040494-6040494 on Build GRCh38
rs779871138 Chr.1: 6040450-6040450 on Build GRCh38
rs771552929 Chr.1: 6040755-6040755 on Build GRCh38
rs1021686391 Chr.1: 6040805-6040805 on Build GRCh38
rs745949762 Chr.1: 6040635-6040635 on Build GRCh38
rs544149386 Chr.1: 6040777-6040777 on Build GRCh38

View Full Product Details