Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6051387-6051804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB2
Entrez Gene ID
8514
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 2
Gene Aliases
AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, KCNA2B, KV-BETA-2
UniGene
Hs.440497
Gene Chromosome Location
Chr.1: 5992298-6101193 on Build GRCh38
Transcripts
NM_001199860.1 NM_001199861.1 NM_001199862.1 NM_001199863.1 NM_003636.3 NM_172130.2 XM_005263514.2 XM_011542321.2 XM_011542322.2 XM_017002618.1 XM_017002619.1 XM_017002620.1 XM_017002621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3786253 c.160C>T p.R54C Chr.1: 6051696-6051696 on Build GRCh38
7327587 c.32G>A p.R11Q Chr.1: 6051568-6051568 on Build GRCh38
7307422 c.122G>A p.R41Q Chr.1: 6051658-6051658 on Build GRCh38
4805971 c.125C>T p.A42V Chr.1: 6051661-6051661 on Build GRCh38
5782970 c.183C>T p.D61D Chr.1: 6051719-6051719 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777063723 Chr.1: 6051500-6051500 on Build GRCh38
rs762932877 Chr.1: 6051753-6051753 on Build GRCh38
rs147130504 Chr.1: 6051550-6051550 on Build GRCh38
rs762334727 Chr.1: 6051760-6051760 on Build GRCh38
rs575930723 Chr.1: 6051472-6051472 on Build GRCh38
rs977047141 Chr.1: 6051775-6051775 on Build GRCh38
rs529702052 Chr.1: 6051641-6051641 on Build GRCh38
rs978607487 Chr.1: 6051739-6051739 on Build GRCh38
rs953498262 Chr.1: 6051573-6051573 on Build GRCh38
rs867008495 Chr.1: 6051736-6051736 on Build GRCh38
rs1006414296 Chr.1: 6051604-6051604 on Build GRCh38
rs915851931 Chr.1: 6051578-6051578 on Build GRCh38
rs1022985862 Chr.1: 6051728-6051728 on Build GRCh38
rs922736222 Chr.1: 6051754-6051754 on Build GRCh38
rs575024397 Chr.1: 6051420-6051420 on Build GRCh38
rs928665371 Chr.1: 6051616-6051616 on Build GRCh38
rs992724431 Chr.1: 6051440-6051440 on Build GRCh38
rs373579322 Chr.1: 6051461-6051461 on Build GRCh38
rs934605189 Chr.1: 6051648-6051648 on Build GRCh38
rs530635823 Chr.1: 6051658-6051658 on Build GRCh38
rs928723090 Chr.1: 6051467-6051467 on Build GRCh38
rs968801038 Chr.1: 6051735-6051735 on Build GRCh38
rs772422878 Chr.1: 6051551-6051551 on Build GRCh38
rs140304262 Chr.1: 6051643-6051643 on Build GRCh38
rs202153711 Chr.1: 6051661-6051661 on Build GRCh38
rs1013948926 Chr.1: 6051701-6051701 on Build GRCh38
rs373219940 Chr.1: 6051705-6051705 on Build GRCh38
rs960001459 Chr.1: 6051720-6051720 on Build GRCh38
rs757693483 Chr.1: 6051471-6051471 on Build GRCh38
rs761654803 Chr.1: 6051615-6051615 on Build GRCh38
rs11589347 Chr.1: 6051697-6051697 on Build GRCh38
rs563117758 Chr.1: 6051649-6051649 on Build GRCh38
rs962047892 Chr.1: 6051433-6051433 on Build GRCh38
rs1016499532 Chr.1: 6051605-6051605 on Build GRCh38
rs564661467 Chr.1: 6051582-6051582 on Build GRCh38
rs1005748225 Chr.1: 6051537-6051537 on Build GRCh38
rs540717541 Chr.1: 6051629-6051629 on Build GRCh38
rs750379162 Chr.1: 6051628-6051628 on Build GRCh38
rs186325427 Chr.1: 6051468-6051468 on Build GRCh38
rs924536381 Chr.1: 6051644-6051644 on Build GRCh38
rs1026407906 Chr.1: 6051568-6051568 on Build GRCh38
rs991466278 Chr.1: 6051726-6051726 on Build GRCh38
rs781152824 Chr.1: 6051611-6051611 on Build GRCh38
rs1035954422 Chr.1: 6051710-6051710 on Build GRCh38
rs747385145 Chr.1: 6051696-6051696 on Build GRCh38
rs769237981 Chr.1: 6051711-6051711 on Build GRCh38
rs781715334 Chr.1: 6051698-6051698 on Build GRCh38
rs775038262 Chr.1: 6051729-6051729 on Build GRCh38
rs1006377045 Chr.1: 6051681-6051681 on Build GRCh38
rs948593484 Chr.1: 6051458-6051458 on Build GRCh38
rs564913084 Chr.1: 6051776-6051776 on Build GRCh38
rs969933391 Chr.1: 6051583-6051583 on Build GRCh38
rs767578775 Chr.1: 6051623-6051623 on Build GRCh38
rs991498904 Chr.1: 6051626-6051626 on Build GRCh38
rs763839795 Chr.1: 6051572-6051572 on Build GRCh38
rs998854387 Chr.1: 6051523-6051523 on Build GRCh38
rs934521476 Chr.1: 6051774-6051774 on Build GRCh38
rs963137597 Chr.1: 6051559-6051559 on Build GRCh38
rs779402966 Chr.1: 6051530-6051530 on Build GRCh38
rs903116426 Chr.1: 6051504-6051504 on Build GRCh38
rs559014790 Chr.1: 6051632-6051632 on Build GRCh38
rs900807078 Chr.1: 6051657-6051657 on Build GRCh38
rs777807905 Chr.1: 6051662-6051662 on Build GRCh38
rs746221121 Chr.1: 6051531-6051531 on Build GRCh38
rs774621032 Chr.1: 6051704-6051704 on Build GRCh38
rs1053545336 Chr.1: 6051473-6051473 on Build GRCh38
rs759053342 Chr.1: 6051501-6051501 on Build GRCh38
rs915805503 Chr.1: 6051455-6051455 on Build GRCh38
rs776169736 Chr.1: 6051595-6051595 on Build GRCh38
rs267598678 Chr.1: 6051713-6051713 on Build GRCh38
rs993237505 Chr.1: 6051567-6051567 on Build GRCh38
rs576750498 Chr.1: 6051593-6051593 on Build GRCh38
rs535005730 Chr.1: 6051764-6051764 on Build GRCh38

View Full Product Details