Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6419643-6420149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HES2
Entrez Gene ID
54626
Gene Name
hes family bHLH transcription factor 2
Gene Aliases
bHLHb40
UniGene
Hs.118727
Gene Chromosome Location
Chr.1: 6415232-6419919 on Build GRCh38
Transcripts
NM_019089.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6433130 c.14G>A p.R5H Chr.1: 6419807-6419807 on Build GRCh38
6317401 c.46-5C>A p.? Chr.1: 6419707-6419707 on Build GRCh38
6433697 c.49C>A p.L17M Chr.1: 6419699-6419699 on Build GRCh38
358063 c.30G>T p.A10A Chr.1: 6419791-6419791 on Build GRCh38
3491725 c.23G>A p.G8E Chr.1: 6419798-6419798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548519359 Chr.1: 6419806-6419806 on Build GRCh38
rs941142900 Chr.1: 6419991-6419991 on Build GRCh38
rs1036757358 Chr.1: 6419875-6419875 on Build GRCh38
rs978798072 Chr.1: 6419693-6419693 on Build GRCh38
rs71299821 Chr.1: 6419973-6419974 on Build GRCh38
rs904677244 Chr.1: 6419901-6419901 on Build GRCh38
rs764249047 Chr.1: 6419760-6419760 on Build GRCh38
rs565651841 Chr.1: 6419894-6419894 on Build GRCh38
rs956068128 Chr.1: 6419932-6419932 on Build GRCh38
rs1002444239 Chr.1: 6420021-6420021 on Build GRCh38
rs767388429 Chr.1: 6419723-6419723 on Build GRCh38
rs576169954 Chr.1: 6420059-6420059 on Build GRCh38
rs576816803 Chr.1: 6419731-6419731 on Build GRCh38
rs972107088 Chr.1: 6419968-6419968 on Build GRCh38
rs747778516 Chr.1: 6419834-6419834 on Build GRCh38
rs954985212 Chr.1: 6420085-6420085 on Build GRCh38
rs896829156 Chr.1: 6419876-6419876 on Build GRCh38
rs773137994 Chr.1: 6419838-6419838 on Build GRCh38
rs554492381 Chr.1: 6419928-6419928 on Build GRCh38
rs958032413 Chr.1: 6420057-6420057 on Build GRCh38
rs988076895 Chr.1: 6420071-6420071 on Build GRCh38
rs12074131 Chr.1: 6419878-6419878 on Build GRCh38
rs750683882 Chr.1: 6419733-6419741 on Build GRCh38
rs1029655109 Chr.1: 6419958-6419958 on Build GRCh38
rs941111397 Chr.1: 6419869-6419869 on Build GRCh38
rs751769139 Chr.1: 6419765-6419765 on Build GRCh38
rs942542362 Chr.1: 6420107-6420107 on Build GRCh38
rs866752443 Chr.1: 6419844-6419844 on Build GRCh38
rs767846365 Chr.1: 6419664-6419665 on Build GRCh38
rs977196137 Chr.1: 6419936-6419936 on Build GRCh38
rs202012466 Chr.1: 6419767-6419767 on Build GRCh38
rs112408380 Chr.1: 6419865-6419865 on Build GRCh38
rs918372937 Chr.1: 6420035-6420035 on Build GRCh38
rs776583914 Chr.1: 6419845-6419845 on Build GRCh38
rs530175882 Chr.1: 6419774-6419774 on Build GRCh38
rs757661625 Chr.1: 6419766-6419766 on Build GRCh38
rs1039638635 Chr.1: 6419893-6419893 on Build GRCh38
rs748767651 Chr.1: 6419821-6419821 on Build GRCh38
rs563885689 Chr.1: 6419830-6419830 on Build GRCh38
rs536988543 Chr.1: 6420095-6420095 on Build GRCh38
rs558425282 Chr.1: 6420120-6420120 on Build GRCh38
rs752724540 Chr.1: 6419736-6419736 on Build GRCh38
rs781689406 Chr.1: 6419664-6419664 on Build GRCh38
rs149736763 Chr.1: 6419759-6419759 on Build GRCh38
rs182995155 Chr.1: 6419706-6419706 on Build GRCh38
rs1046850120 Chr.1: 6419939-6419939 on Build GRCh38
rs970073827 Chr.1: 6419812-6419812 on Build GRCh38
rs780548003 Chr.1: 6419807-6419807 on Build GRCh38
rs773330553 Chr.1: 6419724-6419724 on Build GRCh38
rs761732186 Chr.1: 6419711-6419711 on Build GRCh38
rs909733046 Chr.1: 6419982-6419982 on Build GRCh38
rs968804454 Chr.1: 6419690-6419690 on Build GRCh38
rs942634080 Chr.1: 6419888-6419888 on Build GRCh38
rs985354398 Chr.1: 6419961-6419961 on Build GRCh38
rs925326929 Chr.1: 6419825-6419825 on Build GRCh38
rs1023083897 Chr.1: 6419670-6419670 on Build GRCh38
rs1056563640 Chr.1: 6420047-6420047 on Build GRCh38
rs972515178 Chr.1: 6420007-6420007 on Build GRCh38
rs528360297 Chr.1: 6419973-6419973 on Build GRCh38
rs761576952 Chr.1: 6420079-6420079 on Build GRCh38
rs1044597963 Chr.1: 6420108-6420108 on Build GRCh38
rs746347051 Chr.1: 6419669-6419669 on Build GRCh38
rs751831767 Chr.1: 6419856-6419856 on Build GRCh38
rs1013110848 Chr.1: 6419896-6419896 on Build GRCh38
rs939108750 Chr.1: 6420038-6420038 on Build GRCh38
rs760911003 Chr.1: 6419846-6419847 on Build GRCh38
rs772049404 Chr.1: 6419835-6419835 on Build GRCh38
rs773915521 Chr.1: 6419831-6419831 on Build GRCh38
rs765426557 Chr.1: 6419735-6419735 on Build GRCh38
rs774005281 Chr.1: 6419710-6419710 on Build GRCh38
rs531270068 Chr.1: 6419843-6419843 on Build GRCh38
rs886834405 Chr.1: 6419955-6419955 on Build GRCh38
rs894793617 Chr.1: 6420054-6420054 on Build GRCh38
rs536314530 Chr.1: 6419904-6419904 on Build GRCh38
rs1017979542 Chr.1: 6419688-6419688 on Build GRCh38
rs70981382 Chr.1: 6419977-6419978 on Build GRCh38
rs774855140 Chr.1: 6419694-6419694 on Build GRCh38
rs992695569 Chr.1: 6419738-6419738 on Build GRCh38
rs1023114985 Chr.1: 6419900-6419900 on Build GRCh38
rs188088468 Chr.1: 6419959-6419959 on Build GRCh38
rs571311975 Chr.1: 6419848-6419848 on Build GRCh38
rs932553383 Chr.1: 6419815-6419815 on Build GRCh38
rs1006574225 Chr.1: 6419678-6419678 on Build GRCh38
rs971855690 Chr.1: 6419963-6419963 on Build GRCh38
rs748098479 Chr.1: 6420098-6420098 on Build GRCh38
rs762015094 Chr.1: 6419857-6419857 on Build GRCh38
rs769349251 Chr.1: 6419674-6419674 on Build GRCh38
rs945921426 Chr.1: 6419873-6419873 on Build GRCh38
rs552897481 Chr.1: 6419847-6419847 on Build GRCh38
rs769367479 Chr.1: 6420099-6420099 on Build GRCh38
rs987557547 Chr.1: 6420086-6420086 on Build GRCh38
rs938979264 Chr.1: 6419877-6419877 on Build GRCh38
rs956206149 Chr.1: 6419707-6419707 on Build GRCh38

View Full Product Details