Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53956192-53956721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXC12
Entrez Gene ID
3228
Gene Name
homeobox C12
Gene Aliases
HOC3F, HOX3, HOX3F
UniGene
Hs.381267
Gene Chromosome Location
Chr.12: 53954930-53956566 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4554109 c.611G>A p.G204D Chr.12: 53956328-53956328 on Build GRCh38
6681743 c.764A>T p.D255V Chr.12: 53956481-53956481 on Build GRCh38
6588377 c.840_842delCTT p.F282delF Chr.12: 53956557-53956559 on Build GRCh38
4043131 c.826G>A p.E276K Chr.12: 53956543-53956543 on Build GRCh38
4043130 c.754A>G p.N252D Chr.12: 53956471-53956471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747091631 Chr.12: 53956282-53956282 on Build GRCh38
rs912780796 Chr.12: 53956517-53956517 on Build GRCh38
rs74089826 Chr.12: 53956226-53956226 on Build GRCh38
rs760375489 Chr.12: 53956659-53956659 on Build GRCh38
rs780332219 Chr.12: 53956330-53956330 on Build GRCh38
rs767115188 Chr.12: 53956337-53956337 on Build GRCh38
rs749316497 Chr.12: 53956572-53956572 on Build GRCh38
rs547484499 Chr.12: 53956611-53956611 on Build GRCh38
rs776119815 Chr.12: 53956595-53956595 on Build GRCh38
rs768997409 Chr.12: 53956499-53956499 on Build GRCh38
rs889570582 Chr.12: 53956497-53956497 on Build GRCh38
rs770150435 Chr.12: 53956377-53956377 on Build GRCh38
rs781490572 Chr.12: 53956464-53956464 on Build GRCh38
rs144078525 Chr.12: 53956396-53956396 on Build GRCh38
rs1044188242 Chr.12: 53956318-53956318 on Build GRCh38
rs774946568 Chr.12: 53956556-53956556 on Build GRCh38
rs764960546 Chr.12: 53956608-53956608 on Build GRCh38
rs762634359 Chr.12: 53956618-53956618 on Build GRCh38
rs760134308 Chr.12: 53956428-53956428 on Build GRCh38
rs368956035 Chr.12: 53956447-53956447 on Build GRCh38
rs201053945 Chr.12: 53956582-53956582 on Build GRCh38
rs956060533 Chr.12: 53956637-53956637 on Build GRCh38
rs555869963 Chr.12: 53956237-53956237 on Build GRCh38
rs376505789 Chr.12: 53956310-53956310 on Build GRCh38
rs373624523 Chr.12: 53956543-53956543 on Build GRCh38
rs756665453 Chr.12: 53956568-53956568 on Build GRCh38
rs141144844 Chr.12: 53956687-53956687 on Build GRCh38
rs146914178 Chr.12: 53956486-53956486 on Build GRCh38
rs770848175 Chr.12: 53956590-53956590 on Build GRCh38
rs768610858 Chr.12: 53956397-53956397 on Build GRCh38
rs776042865 Chr.12: 53956679-53956679 on Build GRCh38
rs775806170 Chr.12: 53956387-53956387 on Build GRCh38
rs199571419 Chr.12: 53956531-53956531 on Build GRCh38
rs778955418 Chr.12: 53956579-53956579 on Build GRCh38
rs756601764 Chr.12: 53956698-53956698 on Build GRCh38
rs544968864 Chr.12: 53956289-53956289 on Build GRCh38
rs560346606 Chr.12: 53956361-53956361 on Build GRCh38
rs760578078 Chr.12: 53956535-53956535 on Build GRCh38
rs781116101 Chr.12: 53956422-53956424 on Build GRCh38
rs149417796 Chr.12: 53956550-53956550 on Build GRCh38
rs747603572 Chr.12: 53956365-53956365 on Build GRCh38
rs774203142 Chr.12: 53956500-53956500 on Build GRCh38
rs565133035 Chr.12: 53956536-53956536 on Build GRCh38
rs761458187 Chr.12: 53956508-53956508 on Build GRCh38
rs1028430190 Chr.12: 53956459-53956459 on Build GRCh38
rs753531622 Chr.12: 53956438-53956438 on Build GRCh38
rs757783848 Chr.12: 53956456-53956456 on Build GRCh38
rs768937234 Chr.12: 53956578-53956578 on Build GRCh38
rs779623804 Chr.12: 53956343-53956343 on Build GRCh38
rs772971973 Chr.12: 53956583-53956583 on Build GRCh38
rs12578555 Chr.12: 53956253-53956253 on Build GRCh38
rs780336633 Chr.12: 53956569-53956569 on Build GRCh38
rs548777960 Chr.12: 53956632-53956632 on Build GRCh38
rs751007356 Chr.12: 53956561-53956561 on Build GRCh38
rs752742321 Chr.12: 53956476-53956478 on Build GRCh38
rs941111139 Chr.12: 53956498-53956498 on Build GRCh38
rs576916327 Chr.12: 53956696-53956696 on Build GRCh38
rs1002566040 Chr.12: 53956360-53956360 on Build GRCh38
rs749898210 Chr.12: 53956338-53956338 on Build GRCh38
rs1044521902 Chr.12: 53956596-53956596 on Build GRCh38
rs531309528 Chr.12: 53956273-53956273 on Build GRCh38
rs769953521 Chr.12: 53956473-53956473 on Build GRCh38
rs1049480100 Chr.12: 53956214-53956214 on Build GRCh38
rs186089788 Chr.12: 53956510-53956510 on Build GRCh38
rs745967545 Chr.12: 53956465-53956465 on Build GRCh38
rs140782486 Chr.12: 53956331-53956331 on Build GRCh38
rs755769012 Chr.12: 53956339-53956339 on Build GRCh38
rs377346613 Chr.12: 53956437-53956437 on Build GRCh38
rs374562830 Chr.12: 53956353-53956353 on Build GRCh38
rs748407927 Chr.12: 53956599-53956599 on Build GRCh38
rs761878621 Chr.12: 53956411-53956411 on Build GRCh38
rs77017809 Chr.12: 53956629-53956629 on Build GRCh38
rs34427788 Chr.12: 53956628-53956628 on Build GRCh38
rs771084395 Chr.12: 53956285-53956285 on Build GRCh38
rs994093046 Chr.12: 53956297-53956297 on Build GRCh38
rs751862154 Chr.12: 53956448-53956448 on Build GRCh38
rs934624778 Chr.12: 53956429-53956429 on Build GRCh38
rs748538517 Chr.12: 53956344-53956344 on Build GRCh38
rs76569642 Chr.12: 53956549-53956549 on Build GRCh38
rs200284247 Chr.12: 53956313-53956313 on Build GRCh38
rs746091853 Chr.12: 53956231-53956231 on Build GRCh38
rs146523432 Chr.12: 53956511-53956511 on Build GRCh38
rs754605519 Chr.12: 53956445-53956445 on Build GRCh38
rs138682181 Chr.12: 53956410-53956410 on Build GRCh38
rs749964447 Chr.12: 53956418-53956418 on Build GRCh38
rs749568681 Chr.12: 53956494-53956494 on Build GRCh38
rs1049863396 Chr.12: 53956477-53956477 on Build GRCh38
rs988756836 Chr.12: 53956648-53956648 on Build GRCh38
rs775430122 Chr.12: 53956327-53956327 on Build GRCh38
rs751963240 Chr.12: 53956545-53956545 on Build GRCh38
rs745865686 Chr.12: 53956375-53956375 on Build GRCh38
rs571633593 Chr.12: 53956370-53956370 on Build GRCh38
rs1016595958 Chr.12: 53956248-53956248 on Build GRCh38
rs753594853 Chr.12: 53956525-53956526 on Build GRCh38
rs922877872 Chr.12: 53956658-53956658 on Build GRCh38
rs771326878 Chr.12: 53956369-53956369 on Build GRCh38
rs538889124 Chr.12: 53956666-53956666 on Build GRCh38
rs142687276 Chr.12: 53956479-53956479 on Build GRCh38

View Full Product Details