Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53989436-53989938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXC10
Entrez Gene ID
3226
Gene Name
homeobox C10
Gene Aliases
HOX3I
UniGene
Hs.44276
Gene Chromosome Location
Chr.12: 53985162-53990279 on Build GRCh38
Transcripts
NM_017409.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5481065 c.1027T>C p.*343R Chr.12: 53989444-53989444 on Build GRCh38
3384493 c.1023C>A p.F341L Chr.12: 53989440-53989440 on Build GRCh38
7371434 c.1029A>C p.*343C Chr.12: 53989446-53989446 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752573701 Chr.12: 53989549-53989549 on Build GRCh38
rs774106744 Chr.12: 53989904-53989905 on Build GRCh38
rs754678586 Chr.12: 53989460-53989460 on Build GRCh38
rs984022846 Chr.12: 53989735-53989735 on Build GRCh38
rs1031664394 Chr.12: 53989895-53989895 on Build GRCh38
rs1005636605 Chr.12: 53989540-53989540 on Build GRCh38
rs921078985 Chr.12: 53989728-53989728 on Build GRCh38
rs911678463 Chr.12: 53989563-53989563 on Build GRCh38
rs561577653 Chr.12: 53989650-53989650 on Build GRCh38
rs930565257 Chr.12: 53989692-53989692 on Build GRCh38
rs918912825 Chr.12: 53989879-53989879 on Build GRCh38
rs200363226 Chr.12: 53989519-53989520 on Build GRCh38
rs4237810 Chr.12: 53989716-53989716 on Build GRCh38
rs536936740 Chr.12: 53989825-53989825 on Build GRCh38
rs184950567 Chr.12: 53989473-53989473 on Build GRCh38
rs770222472 Chr.12: 53989491-53989491 on Build GRCh38
rs373631729 Chr.12: 53989457-53989457 on Build GRCh38
rs77499918 Chr.12: 53989679-53989679 on Build GRCh38
rs1049475882 Chr.12: 53989702-53989702 on Build GRCh38
rs904768253 Chr.12: 53989855-53989855 on Build GRCh38
rs566280089 Chr.12: 53989462-53989462 on Build GRCh38
rs749279055 Chr.12: 53989494-53989494 on Build GRCh38
rs776640418 Chr.12: 53989785-53989785 on Build GRCh38
rs538916203 Chr.12: 53989795-53989796 on Build GRCh38
rs752359227 Chr.12: 53989468-53989468 on Build GRCh38
rs781527084 Chr.12: 53989477-53989477 on Build GRCh38
rs181735048 Chr.12: 53989593-53989593 on Build GRCh38
rs965442271 Chr.12: 53989711-53989711 on Build GRCh38
rs1035807358 Chr.12: 53989564-53989564 on Build GRCh38
rs186245023 Chr.12: 53989836-53989836 on Build GRCh38
rs554792945 Chr.12: 53989731-53989731 on Build GRCh38
rs370667773 Chr.12: 53989463-53989463 on Build GRCh38
rs531819179 Chr.12: 53989674-53989674 on Build GRCh38
rs35616568 Chr.12: 53989509-53989509 on Build GRCh38
rs746445222 Chr.12: 53989489-53989489 on Build GRCh38
rs967672222 Chr.12: 53989474-53989474 on Build GRCh38
rs1013146500 Chr.12: 53989610-53989610 on Build GRCh38
rs939086538 Chr.12: 53989796-53989796 on Build GRCh38
rs536539932 Chr.12: 53989595-53989595 on Build GRCh38
rs527624546 Chr.12: 53989795-53989795 on Build GRCh38
rs778387407 Chr.12: 53989466-53989466 on Build GRCh38
rs780124260 Chr.12: 53989492-53989492 on Build GRCh38
rs532742861 Chr.12: 53989732-53989732 on Build GRCh38
rs376953065 Chr.12: 53989839-53989839 on Build GRCh38
rs566420721 Chr.12: 53989750-53989750 on Build GRCh38
rs549008788 Chr.12: 53989816-53989816 on Build GRCh38
rs939994562 Chr.12: 53989775-53989775 on Build GRCh38
rs760685171 Chr.12: 53989454-53989454 on Build GRCh38
rs965393575 Chr.12: 53989908-53989908 on Build GRCh38
rs953518146 Chr.12: 53989803-53989803 on Build GRCh38
rs765992428 Chr.12: 53989455-53989455 on Build GRCh38
rs1037099375 Chr.12: 53989784-53989784 on Build GRCh38
rs189368208 Chr.12: 53989552-53989552 on Build GRCh38
rs571981486 Chr.12: 53989790-53989791 on Build GRCh38
rs1031710777 Chr.12: 53989703-53989703 on Build GRCh38
rs1036745713 Chr.12: 53989519-53989519 on Build GRCh38
rs554485781 Chr.12: 53989521-53989521 on Build GRCh38
rs926127834 Chr.12: 53989481-53989481 on Build GRCh38
rs894621114 Chr.12: 53989827-53989827 on Build GRCh38
rs977827393 Chr.12: 53989651-53989651 on Build GRCh38
rs567415634 Chr.12: 53989821-53989821 on Build GRCh38
rs550996270 Chr.12: 53989745-53989745 on Build GRCh38
rs750436134 Chr.12: 53989453-53989453 on Build GRCh38
rs972254645 Chr.12: 53989837-53989837 on Build GRCh38
rs1034489272 Chr.12: 53989704-53989704 on Build GRCh38
rs939691188 Chr.12: 53989505-53989505 on Build GRCh38
rs1048812575 Chr.12: 53989490-53989490 on Build GRCh38
rs1022306957 Chr.12: 53989637-53989637 on Build GRCh38
rs999148305 Chr.12: 53989890-53989890 on Build GRCh38
rs771170944 Chr.12: 53989779-53989779 on Build GRCh38

View Full Product Details