Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56339338-56339847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL23A
Entrez Gene ID
51561
Gene Name
interleukin 23 subunit alpha
Gene Aliases
IL-23, IL-23A, IL23P19, P19, SGRF
UniGene
Hs.382212
Gene Chromosome Location
Chr.12: 56334159-56340410 on Build GRCh38
Transcripts
NM_016584.2 XM_011538477.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5725731 c.221G>A p.G74E Chr.12: 56339484-56339484 on Build GRCh38
941652 c.356G>A p.S119N Chr.12: 56339785-56339785 on Build GRCh38
941651 c.241G>T p.G81* Chr.12: 56339504-56339504 on Build GRCh38
5644013 c.358C>T p.P120S Chr.12: 56339787-56339787 on Build GRCh38
4043603 c.226G>A p.G76S Chr.12: 56339489-56339489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778563353 Chr.12: 56339446-56339446 on Build GRCh38
rs949479241 Chr.12: 56339693-56339693 on Build GRCh38
rs758481963 Chr.12: 56339660-56339660 on Build GRCh38
rs199656959 Chr.12: 56339536-56339536 on Build GRCh38
rs776113700 Chr.12: 56339668-56339668 on Build GRCh38
rs781334550 Chr.12: 56339385-56339385 on Build GRCh38
rs764509184 Chr.12: 56339555-56339555 on Build GRCh38
rs797016499 Chr.12: 56339611-56339611 on Build GRCh38
rs747841636 Chr.12: 56339646-56339646 on Build GRCh38
rs760945705 Chr.12: 56339483-56339483 on Build GRCh38
rs748666789 Chr.12: 56339798-56339798 on Build GRCh38
rs768984725 Chr.12: 56339673-56339673 on Build GRCh38
rs201309092 Chr.12: 56339435-56339435 on Build GRCh38
rs768621346 Chr.12: 56339442-56339442 on Build GRCh38
rs925540536 Chr.12: 56339623-56339623 on Build GRCh38
rs750410589 Chr.12: 56339406-56339406 on Build GRCh38
rs202153946 Chr.12: 56339495-56339495 on Build GRCh38
rs753906462 Chr.12: 56339720-56339720 on Build GRCh38
rs770394285 Chr.12: 56339519-56339519 on Build GRCh38
rs766514214 Chr.12: 56339741-56339763 on Build GRCh38
rs762044893 Chr.12: 56339685-56339685 on Build GRCh38
rs758098526 Chr.12: 56339653-56339653 on Build GRCh38
rs770482196 Chr.12: 56339663-56339663 on Build GRCh38
rs200803681 Chr.12: 56339789-56339789 on Build GRCh38
rs1000906886 Chr.12: 56339662-56339662 on Build GRCh38
rs767905969 Chr.12: 56339741-56339741 on Build GRCh38
rs750548351 Chr.12: 56339569-56339569 on Build GRCh38
rs200114104 Chr.12: 56339787-56339787 on Build GRCh38
rs774799160 Chr.12: 56339675-56339675 on Build GRCh38
rs754983389 Chr.12: 56339736-56339736 on Build GRCh38
rs763062396 Chr.12: 56339671-56339671 on Build GRCh38
rs779330106 Chr.12: 56339779-56339779 on Build GRCh38
rs746480092 Chr.12: 56339511-56339511 on Build GRCh38
rs773710976 Chr.12: 56339814-56339814 on Build GRCh38
rs976258964 Chr.12: 56339582-56339582 on Build GRCh38
rs921113733 Chr.12: 56339748-56339748 on Build GRCh38
rs150119995 Chr.12: 56339817-56339817 on Build GRCh38
rs965839856 Chr.12: 56339577-56339577 on Build GRCh38
rs762107047 Chr.12: 56339561-56339561 on Build GRCh38
rs770907049 Chr.12: 56339627-56339627 on Build GRCh38
rs568235540 Chr.12: 56339604-56339604 on Build GRCh38
rs113129337 Chr.12: 56339765-56339765 on Build GRCh38
rs778776361 Chr.12: 56339643-56339643 on Build GRCh38
rs921599676 Chr.12: 56339593-56339593 on Build GRCh38
rs749208260 Chr.12: 56339441-56339441 on Build GRCh38
rs200708303 Chr.12: 56339767-56339767 on Build GRCh38
rs201997002 Chr.12: 56339746-56339746 on Build GRCh38
rs751865187 Chr.12: 56339380-56339380 on Build GRCh38
rs772794641 Chr.12: 56339476-56339476 on Build GRCh38
rs773583821 Chr.12: 56339658-56339659 on Build GRCh38
rs774511668 Chr.12: 56339559-56339559 on Build GRCh38
rs1014649599 Chr.12: 56339512-56339512 on Build GRCh38
rs541477832 Chr.12: 56339712-56339712 on Build GRCh38
rs202007974 Chr.12: 56339786-56339786 on Build GRCh38
rs772167575 Chr.12: 56339370-56339370 on Build GRCh38
rs777608040 Chr.12: 56339749-56339749 on Build GRCh38
rs939725713 Chr.12: 56339613-56339613 on Build GRCh38
rs896540334 Chr.12: 56339676-56339678 on Build GRCh38
rs760965618 Chr.12: 56339699-56339699 on Build GRCh38
rs747905636 Chr.12: 56339459-56339459 on Build GRCh38
rs975654194 Chr.12: 56339398-56339398 on Build GRCh38
rs150940553 Chr.12: 56339462-56339462 on Build GRCh38
rs534791691 Chr.12: 56339382-56339382 on Build GRCh38
rs753888175 Chr.12: 56339641-56339641 on Build GRCh38
rs767584027 Chr.12: 56339566-56339566 on Build GRCh38
rs966776168 Chr.12: 56339661-56339661 on Build GRCh38
rs201710057 Chr.12: 56339411-56339411 on Build GRCh38
rs201046773 Chr.12: 56339529-56339529 on Build GRCh38
rs374176852 Chr.12: 56339658-56339658 on Build GRCh38
rs776502340 Chr.12: 56339714-56339714 on Build GRCh38
rs772379441 Chr.12: 56339804-56339804 on Build GRCh38
rs898748434 Chr.12: 56339615-56339615 on Build GRCh38
rs764415400 Chr.12: 56339376-56339376 on Build GRCh38
rs765038229 Chr.12: 56339745-56339745 on Build GRCh38
rs770199764 Chr.12: 56339440-56339440 on Build GRCh38
rs745330561 Chr.12: 56339773-56339773 on Build GRCh38
rs535603776 Chr.12: 56339671-56339671 on Build GRCh38
rs756835874 Chr.12: 56339766-56339766 on Build GRCh38
rs773345315 Chr.12: 56339426-56339426 on Build GRCh38
rs908288141 Chr.12: 56339606-56339606 on Build GRCh38
rs11465746 Chr.12: 56339429-56339429 on Build GRCh38
rs200585151 Chr.12: 56339549-56339549 on Build GRCh38
rs767894389 Chr.12: 56339687-56339687 on Build GRCh38
rs762122707 Chr.12: 56339411-56339414 on Build GRCh38
rs756303469 Chr.12: 56339572-56339572 on Build GRCh38
rs1045586127 Chr.12: 56339721-56339722 on Build GRCh38
rs766545393 Chr.12: 56339640-56339640 on Build GRCh38
rs776021749 Chr.12: 56339535-56339535 on Build GRCh38
rs11171806 Chr.12: 56339747-56339747 on Build GRCh38
rs1050776430 Chr.12: 56339596-56339596 on Build GRCh38
rs896629513 Chr.12: 56339651-56339651 on Build GRCh38
rs201540723 Chr.12: 56339642-56339642 on Build GRCh38
rs200222221 Chr.12: 56339528-56339528 on Build GRCh38
rs200416627 Chr.12: 56339391-56339391 on Build GRCh38
rs1038000570 Chr.12: 56339614-56339614 on Build GRCh38
rs766637414 Chr.12: 56339717-56339717 on Build GRCh38
rs557140087 Chr.12: 56339691-56339691 on Build GRCh38
rs746727343 Chr.12: 56339758-56339758 on Build GRCh38
rs769235407 Chr.12: 56339778-56339778 on Build GRCh38

View Full Product Details