Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103939431-103939915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSP90B1
Entrez Gene ID
7184
Gene Name
heat shock protein 90 beta family member 1
Gene Aliases
ECGP, GP96, GRP94, HEL-S-125m, HEL35, TRA1
UniGene
Hs.192374
Gene Chromosome Location
Chr.12: 103930334-103947930 on Build GRCh38
Transcripts
NM_003299.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4964947 c.1046A>T p.E349V Chr.12: 103939579-103939579 on Build GRCh38
281784 c.1034G>T p.R345I Chr.12: 103939567-103939567 on Build GRCh38
467688 c.1071T>C p.A357A Chr.12: 103939604-103939604 on Build GRCh38
7333721 c.1077C>T p.Y359Y Chr.12: 103939610-103939610 on Build GRCh38
7077518 c.1075T>C p.Y359H Chr.12: 103939608-103939608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053274912 Chr.12: 103939886-103939886 on Build GRCh38
rs753778542 Chr.12: 103939594-103939594 on Build GRCh38
rs373729714 Chr.12: 103939482-103939482 on Build GRCh38
rs767254008 Chr.12: 103939564-103939564 on Build GRCh38
rs934925564 Chr.12: 103939867-103939867 on Build GRCh38
rs17034955 Chr.12: 103939474-103939474 on Build GRCh38
rs773609488 Chr.12: 103939677-103939677 on Build GRCh38
rs879150773 Chr.12: 103939561-103939561 on Build GRCh38
rs748158481 Chr.12: 103939718-103939718 on Build GRCh38
rs759301090 Chr.12: 103939546-103939546 on Build GRCh38
rs549952668 Chr.12: 103939494-103939494 on Build GRCh38
rs776434920 Chr.12: 103939788-103939788 on Build GRCh38
rs199957905 Chr.12: 103939662-103939662 on Build GRCh38
rs781259149 Chr.12: 103939805-103939805 on Build GRCh38
rs748520208 Chr.12: 103939669-103939669 on Build GRCh38
rs747072388 Chr.12: 103939646-103939646 on Build GRCh38
rs768891486 Chr.12: 103939652-103939652 on Build GRCh38
rs980836553 Chr.12: 103939687-103939687 on Build GRCh38
rs553886850 Chr.12: 103939807-103939807 on Build GRCh38
rs758606058 Chr.12: 103939633-103939633 on Build GRCh38
rs1004105012 Chr.12: 103939685-103939685 on Build GRCh38
rs780038723 Chr.12: 103939640-103939640 on Build GRCh38
rs762119595 Chr.12: 103939489-103939489 on Build GRCh38
rs533988701 Chr.12: 103939798-103939798 on Build GRCh38
rs77096967 Chr.12: 103939696-103939696 on Build GRCh38
rs770320858 Chr.12: 103939496-103939496 on Build GRCh38
rs752562529 Chr.12: 103939570-103939570 on Build GRCh38
rs570023365 Chr.12: 103939497-103939497 on Build GRCh38
rs1023202465 Chr.12: 103939612-103939612 on Build GRCh38
rs779002824 Chr.12: 103939475-103939476 on Build GRCh38
rs570502389 Chr.12: 103939854-103939854 on Build GRCh38
rs924979763 Chr.12: 103939692-103939692 on Build GRCh38
rs757032590 Chr.12: 103939462-103939463 on Build GRCh38
rs746079614 Chr.12: 103939495-103939498 on Build GRCh38
rs557357447 Chr.12: 103939694-103939694 on Build GRCh38
rs762534225 Chr.12: 103939509-103939509 on Build GRCh38
rs772144935 Chr.12: 103939731-103939731 on Build GRCh38
rs376600945 Chr.12: 103939645-103939645 on Build GRCh38
rs746156968 Chr.12: 103939858-103939858 on Build GRCh38
rs765860662 Chr.12: 103939528-103939528 on Build GRCh38
rs757217024 Chr.12: 103939607-103939607 on Build GRCh38
rs751073447 Chr.12: 103939540-103939540 on Build GRCh38
rs375594636 Chr.12: 103939591-103939591 on Build GRCh38
rs970290851 Chr.12: 103939801-103939801 on Build GRCh38
rs933142562 Chr.12: 103939742-103939742 on Build GRCh38
rs568328844 Chr.12: 103939779-103939779 on Build GRCh38
rs780256700 Chr.12: 103939634-103939635 on Build GRCh38
rs375562702 Chr.12: 103939461-103939461 on Build GRCh38
rs1051572027 Chr.12: 103939774-103939774 on Build GRCh38
rs770083536 Chr.12: 103939673-103939673 on Build GRCh38
rs755932488 Chr.12: 103939590-103939590 on Build GRCh38

View Full Product Details