Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30202533-30203040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNAL1
Entrez Gene ID
84056
Gene Name
katanin catalytic subunit A1 like 1
Gene Aliases
-
UniGene
Hs.243596
Gene Chromosome Location
Chr.13: 30202626-30307500 on Build GRCh38
Transcripts
NM_001014380.2 NM_032116.4 XM_005266574.2 XM_017020791.1 XM_017020792.1 XM_017020793.1

SNP Detail

SNP ID Chromosome Location
rs916378689 Chr.13: 30202950-30202950 on Build GRCh38
rs941824066 Chr.13: 30202943-30202953 on Build GRCh38
rs2149972 Chr.13: 30202815-30202815 on Build GRCh38
rs145184974 Chr.13: 30202627-30202627 on Build GRCh38
rs187133921 Chr.13: 30202663-30202663 on Build GRCh38
rs939177536 Chr.13: 30202628-30202628 on Build GRCh38
rs534763401 Chr.13: 30202824-30202824 on Build GRCh38
rs990666926 Chr.13: 30202936-30202936 on Build GRCh38
rs993891038 Chr.13: 30202676-30202676 on Build GRCh38
rs540044701 Chr.13: 30202937-30202937 on Build GRCh38
rs879029502 Chr.13: 30202747-30202747 on Build GRCh38
rs767218578 Chr.13: 30202667-30202667 on Build GRCh38
rs759167602 Chr.13: 30202593-30202593 on Build GRCh38
rs549994536 Chr.13: 30202672-30202672 on Build GRCh38
rs1038005451 Chr.13: 30202668-30202668 on Build GRCh38
rs553567339 Chr.13: 30202940-30202940 on Build GRCh38
rs914593038 Chr.13: 30202961-30202961 on Build GRCh38
rs900757958 Chr.13: 30202792-30202792 on Build GRCh38
rs548985794 Chr.13: 30202759-30202759 on Build GRCh38
rs960859235 Chr.13: 30202934-30202934 on Build GRCh38
rs1055481971 Chr.13: 30202657-30202657 on Build GRCh38
rs972424636 Chr.13: 30202751-30202751 on Build GRCh38
rs952996886 Chr.13: 30202642-30202643 on Build GRCh38
rs541985430 Chr.13: 30203001-30203001 on Build GRCh38
rs533005235 Chr.13: 30202594-30202594 on Build GRCh38
rs981455538 Chr.13: 30202854-30202854 on Build GRCh38
rs978603745 Chr.13: 30202912-30202912 on Build GRCh38
rs966674543 Chr.13: 30202846-30202846 on Build GRCh38
rs978171313 Chr.13: 30202601-30202601 on Build GRCh38
rs1050843685 Chr.13: 30202685-30202685 on Build GRCh38
rs557730158 Chr.13: 30202908-30202908 on Build GRCh38
rs35827750 Chr.13: 30202555-30202556 on Build GRCh38
rs765721573 Chr.13: 30203003-30203003 on Build GRCh38
rs890643436 Chr.13: 30202696-30202696 on Build GRCh38
rs566572180 Chr.13: 30202644-30202644 on Build GRCh38
rs961974083 Chr.13: 30202746-30202746 on Build GRCh38
rs528947998 Chr.13: 30202708-30202708 on Build GRCh38
rs1018312183 Chr.13: 30202760-30202760 on Build GRCh38
rs1035096355 Chr.13: 30202932-30202932 on Build GRCh38
rs1011128591 Chr.13: 30202836-30202836 on Build GRCh38
rs902050856 Chr.13: 30202588-30202588 on Build GRCh38
rs927394271 Chr.13: 30202911-30202911 on Build GRCh38
rs915205747 Chr.13: 30202558-30202558 on Build GRCh38
rs562793987 Chr.13: 30202840-30202840 on Build GRCh38
rs573307663 Chr.13: 30202970-30202970 on Build GRCh38
rs571161809 Chr.13: 30202935-30202935 on Build GRCh38

View Full Product Details