Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30205410-30205909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNAL1
Entrez Gene ID
84056
Gene Name
katanin catalytic subunit A1 like 1
Gene Aliases
-
UniGene
Hs.243596
Gene Chromosome Location
Chr.13: 30202626-30307500 on Build GRCh38
Transcripts
NM_001014380.2 NM_032116.4 XM_005266574.2 XM_017020791.1 XM_017020792.1 XM_017020793.1

SNP Detail

SNP ID Chromosome Location
rs886187362 Chr.13: 30205835-30205835 on Build GRCh38
rs965796302 Chr.13: 30205586-30205586 on Build GRCh38
rs956105583 Chr.13: 30205776-30205776 on Build GRCh38
rs976746634 Chr.13: 30205613-30205613 on Build GRCh38
rs1043889306 Chr.13: 30205522-30205522 on Build GRCh38
rs1028565083 Chr.13: 30205872-30205872 on Build GRCh38
rs895500500 Chr.13: 30205450-30205450 on Build GRCh38
rs112323204 Chr.13: 30205833-30205833 on Build GRCh38
rs796164666 Chr.13: 30205613-30205614 on Build GRCh38
rs1004897437 Chr.13: 30205580-30205580 on Build GRCh38
rs878910890 Chr.13: 30205754-30205755 on Build GRCh38
rs190003256 Chr.13: 30205727-30205727 on Build GRCh38
rs34674202 Chr.13: 30205636-30205636 on Build GRCh38
rs567669260 Chr.13: 30205734-30205734 on Build GRCh38
rs759740433 Chr.13: 30205711-30205712 on Build GRCh38
rs796174197 Chr.13: 30205758-30205758 on Build GRCh38
rs879447828 Chr.13: 30205756-30205756 on Build GRCh38
rs146509188 Chr.13: 30205750-30205751 on Build GRCh38
rs748663473 Chr.13: 30205631-30205635 on Build GRCh38
rs1048781521 Chr.13: 30205709-30205709 on Build GRCh38
rs1044161198 Chr.13: 30205502-30205502 on Build GRCh38
rs895383335 Chr.13: 30205453-30205453 on Build GRCh38
rs73163456 Chr.13: 30205637-30205637 on Build GRCh38
rs979964091 Chr.13: 30205801-30205801 on Build GRCh38
rs934498242 Chr.13: 30205667-30205667 on Build GRCh38
rs181769499 Chr.13: 30205871-30205871 on Build GRCh38
rs905505769 Chr.13: 30205700-30205700 on Build GRCh38
rs9508677 Chr.13: 30205857-30205857 on Build GRCh38
rs797015785 Chr.13: 30205546-30205546 on Build GRCh38
rs397836237 Chr.13: 30205788-30205789 on Build GRCh38
rs930299613 Chr.13: 30205821-30205821 on Build GRCh38
rs1017982645 Chr.13: 30205629-30205629 on Build GRCh38
rs148459455 Chr.13: 30205631-30205632 on Build GRCh38
rs921227843 Chr.13: 30205615-30205615 on Build GRCh38
rs11620056 Chr.13: 30205682-30205682 on Build GRCh38
rs536213390 Chr.13: 30205792-30205792 on Build GRCh38
rs1030771155 Chr.13: 30205565-30205565 on Build GRCh38
rs55662051 Chr.13: 30205643-30205648 on Build GRCh38
rs143277319 Chr.13: 30205751-30205752 on Build GRCh38
rs1044372078 Chr.13: 30205750-30205757 on Build GRCh38
rs948326830 Chr.13: 30205750-30205751 on Build GRCh38
rs939366446 Chr.13: 30205439-30205439 on Build GRCh38
rs865774167 Chr.13: 30205760-30205760 on Build GRCh38
rs1013913633 Chr.13: 30205458-30205458 on Build GRCh38
rs201006575 Chr.13: 30205632-30205637 on Build GRCh38
rs1017953241 Chr.13: 30205847-30205847 on Build GRCh38
rs9508676 Chr.13: 30205855-30205855 on Build GRCh38
rs201837411 Chr.13: 30205640-30205643 on Build GRCh38
rs994510853 Chr.13: 30205542-30205542 on Build GRCh38
rs560912166 Chr.13: 30205443-30205443 on Build GRCh38
rs2039708 Chr.13: 30205632-30205632 on Build GRCh38
rs373439850 Chr.13: 30205701-30205701 on Build GRCh38
rs879646704 Chr.13: 30205545-30205545 on Build GRCh38
rs998405741 Chr.13: 30205754-30205754 on Build GRCh38
rs1037211516 Chr.13: 30205846-30205846 on Build GRCh38
rs374515272 Chr.13: 30205634-30205637 on Build GRCh38
rs117991635 Chr.13: 30205852-30205852 on Build GRCh38
rs755644121 Chr.13: 30205853-30205854 on Build GRCh38
rs1033089186 Chr.13: 30205762-30205762 on Build GRCh38
rs113722190 Chr.13: 30205633-30205638 on Build GRCh38
rs974438478 Chr.13: 30205680-30205680 on Build GRCh38
rs386769583 Chr.13: 30205637-30205643 on Build GRCh38
rs989168854 Chr.13: 30205675-30205675 on Build GRCh38
rs747778315 Chr.13: 30205637-30205638 on Build GRCh38
rs866037092 Chr.13: 30205643-30205643 on Build GRCh38
rs114243171 Chr.13: 30205795-30205795 on Build GRCh38
rs202216815 Chr.13: 30205636-30205637 on Build GRCh38
rs761701257 Chr.13: 30205750-30205759 on Build GRCh38
rs1012942380 Chr.13: 30205520-30205520 on Build GRCh38
rs112146305 Chr.13: 30205845-30205845 on Build GRCh38
rs531790320 Chr.13: 30205656-30205656 on Build GRCh38
rs151034112 Chr.13: 30205558-30205558 on Build GRCh38
rs777009591 Chr.13: 30205636-30205643 on Build GRCh38
rs562835123 Chr.13: 30205651-30205651 on Build GRCh38
rs1015835852 Chr.13: 30205585-30205585 on Build GRCh38
rs963988959 Chr.13: 30205638-30205647 on Build GRCh38
rs34617448 Chr.13: 30205638-30205638 on Build GRCh38
rs890866986 Chr.13: 30205832-30205835 on Build GRCh38
rs769563729 Chr.13: 30205552-30205552 on Build GRCh38
rs71093027 Chr.13: 30205632-30205633 on Build GRCh38
rs368040449 Chr.13: 30205674-30205675 on Build GRCh38
rs770799643 Chr.13: 30205753-30205754 on Build GRCh38
rs777717203 Chr.13: 30205750-30205753 on Build GRCh38
rs904056887 Chr.13: 30205533-30205542 on Build GRCh38
rs940398150 Chr.13: 30205836-30205836 on Build GRCh38
rs1044046237 Chr.13: 30205693-30205693 on Build GRCh38
rs78978705 Chr.13: 30205752-30205752 on Build GRCh38
rs988661568 Chr.13: 30205781-30205781 on Build GRCh38
rs907848088 Chr.13: 30205436-30205436 on Build GRCh38
rs3041555 Chr.13: 30205760-30205761 on Build GRCh38
rs952889322 Chr.13: 30205562-30205562 on Build GRCh38
rs114829192 Chr.13: 30205731-30205731 on Build GRCh38
rs905621171 Chr.13: 30205521-30205521 on Build GRCh38
rs767408433 Chr.13: 30205751-30205754 on Build GRCh38
rs200295479 Chr.13: 30205638-30205638 on Build GRCh38
rs868864899 Chr.13: 30205618-30205618 on Build GRCh38
rs926986351 Chr.13: 30205803-30205803 on Build GRCh38
rs916602266 Chr.13: 30205745-30205745 on Build GRCh38
rs141266150 Chr.13: 30205692-30205692 on Build GRCh38
rs71209443 Chr.13: 30205632-30205638 on Build GRCh38
rs375460166 Chr.13: 30205751-30205760 on Build GRCh38

View Full Product Details