Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30205903-30206447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNAL1
Entrez Gene ID
84056
Gene Name
katanin catalytic subunit A1 like 1
Gene Aliases
-
UniGene
Hs.243596
Gene Chromosome Location
Chr.13: 30202626-30307500 on Build GRCh38
Transcripts
NM_001014380.2 NM_032116.4 XM_005266574.2 XM_017020791.1 XM_017020792.1 XM_017020793.1

SNP Detail

SNP ID Chromosome Location
rs1024151291 Chr.13: 30206310-30206310 on Build GRCh38
rs57329263 Chr.13: 30206260-30206260 on Build GRCh38
rs989272203 Chr.13: 30206001-30206001 on Build GRCh38
rs186349759 Chr.13: 30206036-30206036 on Build GRCh38
rs145138239 Chr.13: 30206058-30206058 on Build GRCh38
rs557778714 Chr.13: 30206371-30206371 on Build GRCh38
rs545228272 Chr.13: 30206111-30206111 on Build GRCh38
rs943731976 Chr.13: 30206181-30206181 on Build GRCh38
rs1039869326 Chr.13: 30206189-30206189 on Build GRCh38
rs878885915 Chr.13: 30205946-30205946 on Build GRCh38
rs940466208 Chr.13: 30206088-30206088 on Build GRCh38
rs1010343340 Chr.13: 30206240-30206240 on Build GRCh38
rs10161651 Chr.13: 30206321-30206321 on Build GRCh38
rs574383190 Chr.13: 30206057-30206057 on Build GRCh38
rs571788849 Chr.13: 30206084-30206084 on Build GRCh38
rs538894799 Chr.13: 30206141-30206141 on Build GRCh38
rs770469967 Chr.13: 30206089-30206089 on Build GRCh38
rs541141860 Chr.13: 30206010-30206010 on Build GRCh38
rs891341941 Chr.13: 30206161-30206161 on Build GRCh38
rs558863236 Chr.13: 30206387-30206387 on Build GRCh38
rs982559523 Chr.13: 30205944-30205944 on Build GRCh38
rs1035484677 Chr.13: 30206292-30206292 on Build GRCh38
rs1054473988 Chr.13: 30206199-30206199 on Build GRCh38
rs540358214 Chr.13: 30206230-30206230 on Build GRCh38
rs1004117223 Chr.13: 30206272-30206272 on Build GRCh38
rs995597595 Chr.13: 30206229-30206229 on Build GRCh38
rs971187358 Chr.13: 30206018-30206018 on Build GRCh38
rs1035514081 Chr.13: 30205982-30205982 on Build GRCh38
rs139021817 Chr.13: 30206080-30206080 on Build GRCh38
rs956156225 Chr.13: 30205988-30205988 on Build GRCh38
rs956941319 Chr.13: 30206353-30206353 on Build GRCh38
rs572074940 Chr.13: 30206009-30206009 on Build GRCh38
rs547880553 Chr.13: 30206162-30206162 on Build GRCh38
rs1026674184 Chr.13: 30206255-30206255 on Build GRCh38
rs59018192 Chr.13: 30206316-30206317 on Build GRCh38
rs934137377 Chr.13: 30206108-30206108 on Build GRCh38
rs560679786 Chr.13: 30206251-30206251 on Build GRCh38
rs951973903 Chr.13: 30206381-30206381 on Build GRCh38
rs988397222 Chr.13: 30206373-30206373 on Build GRCh38
rs73443641 Chr.13: 30206350-30206350 on Build GRCh38
rs891766553 Chr.13: 30206223-30206223 on Build GRCh38
rs984479274 Chr.13: 30206076-30206076 on Build GRCh38
rs116321103 Chr.13: 30206314-30206314 on Build GRCh38
rs146285301 Chr.13: 30206293-30206294 on Build GRCh38
rs10161646 Chr.13: 30206150-30206150 on Build GRCh38
rs912704216 Chr.13: 30206391-30206391 on Build GRCh38
rs370336610 Chr.13: 30206206-30206206 on Build GRCh38
rs922792819 Chr.13: 30206101-30206101 on Build GRCh38
rs951754136 Chr.13: 30206065-30206065 on Build GRCh38
rs565161021 Chr.13: 30206135-30206135 on Build GRCh38
rs1013589124 Chr.13: 30206307-30206307 on Build GRCh38
rs549748835 Chr.13: 30205978-30205983 on Build GRCh38
rs959867523 Chr.13: 30206298-30206298 on Build GRCh38
rs948056996 Chr.13: 30206062-30206062 on Build GRCh38
rs890042401 Chr.13: 30206142-30206142 on Build GRCh38
rs558493120 Chr.13: 30206094-30206094 on Build GRCh38

View Full Product Details