Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30207111-30207623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNAL1
Entrez Gene ID
84056
Gene Name
katanin catalytic subunit A1 like 1
Gene Aliases
-
UniGene
Hs.243596
Gene Chromosome Location
Chr.13: 30202626-30307500 on Build GRCh38
Transcripts
NM_001014380.2 NM_032116.4 XM_005266574.2 XM_017020791.1 XM_017020792.1 XM_017020793.1

SNP Detail

SNP ID Chromosome Location
rs888690106 Chr.13: 30207356-30207356 on Build GRCh38
rs143978228 Chr.13: 30207299-30207299 on Build GRCh38
rs1041156535 Chr.13: 30207251-30207251 on Build GRCh38
rs769621032 Chr.13: 30207439-30207439 on Build GRCh38
rs879208278 Chr.13: 30207158-30207158 on Build GRCh38
rs1036615922 Chr.13: 30207208-30207208 on Build GRCh38
rs868634684 Chr.13: 30207483-30207483 on Build GRCh38
rs888818197 Chr.13: 30207229-30207229 on Build GRCh38
rs1020892595 Chr.13: 30207330-30207330 on Build GRCh38
rs148598743 Chr.13: 30207353-30207353 on Build GRCh38
rs937330486 Chr.13: 30207179-30207179 on Build GRCh38
rs991454885 Chr.13: 30207447-30207447 on Build GRCh38
rs1015676716 Chr.13: 30207470-30207470 on Build GRCh38
rs527799036 Chr.13: 30207218-30207218 on Build GRCh38
rs3825533 Chr.13: 30207505-30207505 on Build GRCh38
rs958035795 Chr.13: 30207306-30207306 on Build GRCh38
rs1055844303 Chr.13: 30207186-30207186 on Build GRCh38
rs138291048 Chr.13: 30207172-30207172 on Build GRCh38
rs897633692 Chr.13: 30207210-30207210 on Build GRCh38
rs928664786 Chr.13: 30207148-30207148 on Build GRCh38
rs866120580 Chr.13: 30207494-30207494 on Build GRCh38
rs868273915 Chr.13: 30207589-30207589 on Build GRCh38
rs1053876700 Chr.13: 30207562-30207562 on Build GRCh38
rs772835112 Chr.13: 30207446-30207446 on Build GRCh38
rs149481473 Chr.13: 30207195-30207195 on Build GRCh38
rs997458087 Chr.13: 30207225-30207225 on Build GRCh38
rs958907227 Chr.13: 30207442-30207442 on Build GRCh38
rs758519075 Chr.13: 30207228-30207228 on Build GRCh38
rs967556126 Chr.13: 30207344-30207344 on Build GRCh38
rs548389861 Chr.13: 30207545-30207545 on Build GRCh38
rs1024078913 Chr.13: 30207511-30207511 on Build GRCh38
rs1036429931 Chr.13: 30207504-30207504 on Build GRCh38
rs563731425 Chr.13: 30207363-30207363 on Build GRCh38
rs935947336 Chr.13: 30207189-30207189 on Build GRCh38
rs532552098 Chr.13: 30207393-30207393 on Build GRCh38
rs988727612 Chr.13: 30207588-30207588 on Build GRCh38
rs981689896 Chr.13: 30207431-30207431 on Build GRCh38
rs1007448586 Chr.13: 30207559-30207559 on Build GRCh38
rs996876143 Chr.13: 30207291-30207291 on Build GRCh38
rs559250241 Chr.13: 30207142-30207142 on Build GRCh38
rs552283762 Chr.13: 30207456-30207456 on Build GRCh38
rs893451446 Chr.13: 30207564-30207564 on Build GRCh38
rs979781554 Chr.13: 30207526-30207526 on Build GRCh38
rs780668159 Chr.13: 30207193-30207193 on Build GRCh38
rs186795070 Chr.13: 30207524-30207524 on Build GRCh38
rs183006904 Chr.13: 30207288-30207288 on Build GRCh38
rs568265498 Chr.13: 30207499-30207505 on Build GRCh38
rs933131597 Chr.13: 30207515-30207515 on Build GRCh38
rs925875818 Chr.13: 30207174-30207174 on Build GRCh38
rs1009289548 Chr.13: 30207314-30207314 on Build GRCh38
rs758651918 Chr.13: 30207331-30207332 on Build GRCh38
rs909184192 Chr.13: 30207471-30207471 on Build GRCh38
rs1051722352 Chr.13: 30207538-30207538 on Build GRCh38
rs980406033 Chr.13: 30207164-30207164 on Build GRCh38

View Full Product Details