Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30210224-30210727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNAL1
Entrez Gene ID
84056
Gene Name
katanin catalytic subunit A1 like 1
Gene Aliases
-
UniGene
Hs.243596
Gene Chromosome Location
Chr.13: 30202626-30307500 on Build GRCh38
Transcripts
NM_001014380.2 NM_032116.4 XM_005266574.2 XM_017020791.1 XM_017020792.1 XM_017020793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6690273 c.1253C>T p.A418V Chr.13: 30210337-30210337 on Build GRCh38
254752 c.1204G>T p.D402Y Chr.13: 30210386-30210386 on Build GRCh38
1366309 c.1192G>A p.E398K Chr.13: 30210398-30210398 on Build GRCh38
5512975 c.1274+10delT p.? Chr.13: 30210306-30210306 on Build GRCh38
257198 c.1183C>T p.R395C Chr.13: 30210407-30210407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052792728 Chr.13: 30210520-30210520 on Build GRCh38
rs191215967 Chr.13: 30210634-30210634 on Build GRCh38
rs186412462 Chr.13: 30210534-30210534 on Build GRCh38
rs892672418 Chr.13: 30210526-30210526 on Build GRCh38
rs775912042 Chr.13: 30210313-30210313 on Build GRCh38
rs757865403 Chr.13: 30210471-30210471 on Build GRCh38
rs373173743 Chr.13: 30210490-30210490 on Build GRCh38
rs869051916 Chr.13: 30210668-30210670 on Build GRCh38
rs201041051 Chr.13: 30210666-30210666 on Build GRCh38
rs867510649 Chr.13: 30210667-30210667 on Build GRCh38
rs748622013 Chr.13: 30210554-30210554 on Build GRCh38
rs1002844384 Chr.13: 30210606-30210606 on Build GRCh38
rs142681398 Chr.13: 30210269-30210269 on Build GRCh38
rs569043278 Chr.13: 30210382-30210382 on Build GRCh38
rs886615310 Chr.13: 30210528-30210528 on Build GRCh38
rs762231119 Chr.13: 30210431-30210431 on Build GRCh38
rs33951005 Chr.13: 30210651-30210651 on Build GRCh38
rs952677228 Chr.13: 30210669-30210669 on Build GRCh38
rs373747807 Chr.13: 30210464-30210464 on Build GRCh38
rs201513671 Chr.13: 30210398-30210398 on Build GRCh38
rs753628719 Chr.13: 30210363-30210363 on Build GRCh38
rs11617724 Chr.13: 30210429-30210429 on Build GRCh38
rs1022267950 Chr.13: 30210693-30210693 on Build GRCh38
rs75470540 Chr.13: 30210300-30210300 on Build GRCh38
rs56120845 Chr.13: 30210283-30210283 on Build GRCh38
rs780668122 Chr.13: 30210396-30210396 on Build GRCh38
rs867126984 Chr.13: 30210591-30210591 on Build GRCh38
rs905456038 Chr.13: 30210566-30210566 on Build GRCh38
rs751385512 Chr.13: 30210478-30210478 on Build GRCh38
rs763542806 Chr.13: 30210320-30210320 on Build GRCh38
rs576369511 Chr.13: 30210650-30210650 on Build GRCh38
rs796831438 Chr.13: 30210651-30210652 on Build GRCh38
rs757042026 Chr.13: 30210485-30210485 on Build GRCh38
rs1027009964 Chr.13: 30210623-30210623 on Build GRCh38
rs371627792 Chr.13: 30210324-30210324 on Build GRCh38
rs1035202486 Chr.13: 30210555-30210555 on Build GRCh38
rs760539110 Chr.13: 30210651-30210654 on Build GRCh38
rs535686214 Chr.13: 30210512-30210512 on Build GRCh38
rs894921055 Chr.13: 30210632-30210632 on Build GRCh38
rs769228528 Chr.13: 30210422-30210422 on Build GRCh38
rs397851638 Chr.13: 30210653-30210653 on Build GRCh38
rs762924589 Chr.13: 30210305-30210306 on Build GRCh38
rs200409413 Chr.13: 30210465-30210465 on Build GRCh38
rs766457095 Chr.13: 30210378-30210378 on Build GRCh38
rs773147478 Chr.13: 30210650-30210653 on Build GRCh38
rs760709299 Chr.13: 30210650-30210652 on Build GRCh38
rs556373334 Chr.13: 30210565-30210565 on Build GRCh38
rs758325851 Chr.13: 30210387-30210387 on Build GRCh38
rs772455699 Chr.13: 30210436-30210436 on Build GRCh38
rs183166828 Chr.13: 30210389-30210389 on Build GRCh38
rs932528264 Chr.13: 30210500-30210500 on Build GRCh38
rs9508680 Chr.13: 30210392-30210392 on Build GRCh38
rs998534374 Chr.13: 30210260-30210260 on Build GRCh38
rs751295844 Chr.13: 30210395-30210395 on Build GRCh38
rs1011903802 Chr.13: 30210649-30210649 on Build GRCh38
rs774790163 Chr.13: 30210355-30210355 on Build GRCh38
rs768701583 Chr.13: 30210652-30210654 on Build GRCh38
rs201290346 Chr.13: 30210651-30210653 on Build GRCh38
rs116191664 Chr.13: 30210462-30210462 on Build GRCh38
rs762143192 Chr.13: 30210362-30210362 on Build GRCh38
rs934436971 Chr.13: 30210515-30210515 on Build GRCh38
rs769693464 Chr.13: 30210274-30210275 on Build GRCh38
rs1013707046 Chr.13: 30210532-30210532 on Build GRCh38
rs779612220 Chr.13: 30210399-30210399 on Build GRCh38
rs780766683 Chr.13: 30210288-30210288 on Build GRCh38
rs377235230 Chr.13: 30210468-30210468 on Build GRCh38
rs57042245 Chr.13: 30210670-30210670 on Build GRCh38
rs1042743704 Chr.13: 30210509-30210509 on Build GRCh38
rs745314131 Chr.13: 30210297-30210297 on Build GRCh38
rs549156683 Chr.13: 30210368-30210368 on Build GRCh38
rs977909400 Chr.13: 30210402-30210402 on Build GRCh38
rs376148405 Chr.13: 30210415-30210415 on Build GRCh38
rs756926320 Chr.13: 30210367-30210367 on Build GRCh38
rs762016994 Chr.13: 30210653-30210654 on Build GRCh38
rs767331891 Chr.13: 30210486-30210486 on Build GRCh38
rs9508681 Chr.13: 30210417-30210417 on Build GRCh38
rs368277485 Chr.13: 30210270-30210270 on Build GRCh38
rs772987395 Chr.13: 30210289-30210290 on Build GRCh38
rs749819072 Chr.13: 30210406-30210406 on Build GRCh38

View Full Product Details