Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45136522-45137067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F2
Entrez Gene ID
2963
Gene Name
general transcription factor IIF subunit 2
Gene Aliases
BTF4, RAP30, TF2F2, TFIIF
UniGene
Hs.654582
Gene Chromosome Location
Chr.13: 45119830-45284105 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_011535053.2 XM_017020550.1 XM_017020551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1662092 c.94G>C p.A32P Chr.13: 45136760-45136760 on Build GRCh38
6676700 c.131G>A p.R44Q Chr.13: 45136797-45136797 on Build GRCh38
6032740 c.130C>T p.R44W Chr.13: 45136796-45136796 on Build GRCh38
6676698 c.87G>A p.Q29Q Chr.13: 45136753-45136753 on Build GRCh38
244840 c.100G>A p.A34T Chr.13: 45136766-45136766 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs936259854 Chr.13: 45136685-45136685 on Build GRCh38
rs746107381 Chr.13: 45136618-45136618 on Build GRCh38
rs775071878 Chr.13: 45136840-45136840 on Build GRCh38
rs531682656 Chr.13: 45136991-45136991 on Build GRCh38
rs958672376 Chr.13: 45136584-45136584 on Build GRCh38
rs544686461 Chr.13: 45136797-45136797 on Build GRCh38
rs752227528 Chr.13: 45136789-45136790 on Build GRCh38
rs745488345 Chr.13: 45136836-45136836 on Build GRCh38
rs983179210 Chr.13: 45136879-45136879 on Build GRCh38
rs772623280 Chr.13: 45136688-45136688 on Build GRCh38
rs542389768 Chr.13: 45136863-45136863 on Build GRCh38
rs764129745 Chr.13: 45136731-45136731 on Build GRCh38
rs776169934 Chr.13: 45136702-45136702 on Build GRCh38
rs755274273 Chr.13: 45136770-45136770 on Build GRCh38
rs535746137 Chr.13: 45136789-45136789 on Build GRCh38
rs200368894 Chr.13: 45136744-45136744 on Build GRCh38
rs777143669 Chr.13: 45137006-45137006 on Build GRCh38
rs747342377 Chr.13: 45136956-45136956 on Build GRCh38
rs760156399 Chr.13: 45136832-45136832 on Build GRCh38
rs773564996 Chr.13: 45136689-45136689 on Build GRCh38
rs543943162 Chr.13: 45136993-45136993 on Build GRCh38
rs569900224 Chr.13: 45136576-45136576 on Build GRCh38
rs565237223 Chr.13: 45137035-45137035 on Build GRCh38
rs899129608 Chr.13: 45136553-45136553 on Build GRCh38
rs145466997 Chr.13: 45136753-45136753 on Build GRCh38
rs139949632 Chr.13: 45136579-45136579 on Build GRCh38
rs944101976 Chr.13: 45136948-45136948 on Build GRCh38
rs931768340 Chr.13: 45136987-45136987 on Build GRCh38
rs559978518 Chr.13: 45136812-45136812 on Build GRCh38
rs572029316 Chr.13: 45136842-45136842 on Build GRCh38
rs1014386077 Chr.13: 45137033-45137033 on Build GRCh38
rs763790620 Chr.13: 45136836-45136838 on Build GRCh38
rs185135542 Chr.13: 45136610-45136610 on Build GRCh38
rs750130020 Chr.13: 45136772-45136772 on Build GRCh38
rs769550274 Chr.13: 45136839-45136839 on Build GRCh38
rs921361971 Chr.13: 45136984-45136984 on Build GRCh38
rs375261432 Chr.13: 45136798-45136798 on Build GRCh38
rs892197498 Chr.13: 45136647-45136647 on Build GRCh38
rs571654691 Chr.13: 45136591-45136591 on Build GRCh38
rs770517057 Chr.13: 45136699-45136699 on Build GRCh38
rs747381194 Chr.13: 45136691-45136691 on Build GRCh38
rs202043550 Chr.13: 45136701-45136701 on Build GRCh38
rs1050290799 Chr.13: 45136989-45136989 on Build GRCh38
rs953029178 Chr.13: 45136867-45136867 on Build GRCh38
rs370193338 Chr.13: 45136732-45136732 on Build GRCh38
rs1024350838 Chr.13: 45136608-45136608 on Build GRCh38
rs759047531 Chr.13: 45136711-45136711 on Build GRCh38
rs936630794 Chr.13: 45136994-45136994 on Build GRCh38
rs369599150 Chr.13: 45136687-45136687 on Build GRCh38
rs758838760 Chr.13: 45136792-45136792 on Build GRCh38
rs111387853 Chr.13: 45136760-45136760 on Build GRCh38
rs190042066 Chr.13: 45136677-45136677 on Build GRCh38
rs115909751 Chr.13: 45136723-45136723 on Build GRCh38
rs771231031 Chr.13: 45136819-45136819 on Build GRCh38
rs757139282 Chr.13: 45136733-45136733 on Build GRCh38
rs887228193 Chr.13: 45136785-45136785 on Build GRCh38
rs762976872 Chr.13: 45136729-45136729 on Build GRCh38
rs753972000 Chr.13: 45136756-45136756 on Build GRCh38
rs9534041 Chr.13: 45136613-45136613 on Build GRCh38
rs373769946 Chr.13: 45136697-45136697 on Build GRCh38
rs768967090 Chr.13: 45136974-45136974 on Build GRCh38
rs1051198706 Chr.13: 45136778-45136778 on Build GRCh38
rs1014788450 Chr.13: 45136550-45136550 on Build GRCh38
rs3803244 Chr.13: 45136630-45136630 on Build GRCh38
rs778843129 Chr.13: 45136776-45136776 on Build GRCh38
rs778270263 Chr.13: 45136796-45136796 on Build GRCh38
rs781650780 Chr.13: 45136834-45136834 on Build GRCh38
rs566387212 Chr.13: 45136622-45136622 on Build GRCh38
rs764623115 Chr.13: 45136719-45136719 on Build GRCh38
rs908566744 Chr.13: 45136924-45136924 on Build GRCh38
rs560783011 Chr.13: 45136934-45136934 on Build GRCh38
rs561017789 Chr.13: 45136575-45136575 on Build GRCh38
rs199678448 Chr.13: 45136765-45136765 on Build GRCh38
rs1034575283 Chr.13: 45136876-45136876 on Build GRCh38
rs762562456 Chr.13: 45136841-45136841 on Build GRCh38

View Full Product Details