Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45151323-45151847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F2
Entrez Gene ID
2963
Gene Name
general transcription factor IIF subunit 2
Gene Aliases
BTF4, RAP30, TF2F2, TFIIF
UniGene
Hs.654582
Gene Chromosome Location
Chr.13: 45119830-45284105 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_011535053.2 XM_017020550.1 XM_017020551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6447938 c.230C>T p.A77V Chr.13: 45151757-45151757 on Build GRCh38
6358671 c.294G>C p.E98D Chr.13: 45151821-45151821 on Build GRCh38
1941494 c.252C>T p.V84V Chr.13: 45151779-45151779 on Build GRCh38
7370490 c.203T>C p.I68T Chr.13: 45151730-45151730 on Build GRCh38
947634 c.193A>T p.I65F Chr.13: 45151720-45151720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932806458 Chr.13: 45151556-45151556 on Build GRCh38
rs750816860 Chr.13: 45151637-45151637 on Build GRCh38
rs751698945 Chr.13: 45151726-45151726 on Build GRCh38
rs938988070 Chr.13: 45151568-45151568 on Build GRCh38
rs781727810 Chr.13: 45151750-45151750 on Build GRCh38
rs201627484 Chr.13: 45151687-45151687 on Build GRCh38
rs1038124995 Chr.13: 45151619-45151619 on Build GRCh38
rs761641975 Chr.13: 45151657-45151657 on Build GRCh38
rs971572654 Chr.13: 45151430-45151430 on Build GRCh38
rs199984320 Chr.13: 45151682-45151682 on Build GRCh38
rs775305457 Chr.13: 45151661-45151661 on Build GRCh38
rs151182400 Chr.13: 45151724-45151724 on Build GRCh38
rs140156787 Chr.13: 45151639-45151639 on Build GRCh38
rs957661474 Chr.13: 45151473-45151473 on Build GRCh38
rs143003112 Chr.13: 45151692-45151692 on Build GRCh38
rs990342524 Chr.13: 45151512-45151512 on Build GRCh38
rs1050366009 Chr.13: 45151594-45151594 on Build GRCh38
rs1048960402 Chr.13: 45151709-45151709 on Build GRCh38
rs140189009 Chr.13: 45151783-45151783 on Build GRCh38
rs770682484 Chr.13: 45151803-45151803 on Build GRCh38
rs1022686314 Chr.13: 45151417-45151417 on Build GRCh38
rs994328823 Chr.13: 45151627-45151628 on Build GRCh38
rs188696761 Chr.13: 45151438-45151438 on Build GRCh38
rs990264218 Chr.13: 45151490-45151490 on Build GRCh38
rs957403411 Chr.13: 45151707-45151707 on Build GRCh38
rs1013196229 Chr.13: 45151414-45151414 on Build GRCh38
rs771848287 Chr.13: 45151789-45151789 on Build GRCh38
rs945195140 Chr.13: 45151532-45151532 on Build GRCh38
rs79127508 Chr.13: 45151432-45151432 on Build GRCh38
rs897325848 Chr.13: 45151624-45151625 on Build GRCh38
rs993406044 Chr.13: 45151368-45151368 on Build GRCh38
rs978002296 Chr.13: 45151538-45151538 on Build GRCh38
rs376097840 Chr.13: 45151676-45151676 on Build GRCh38
rs780601888 Chr.13: 45151771-45151771 on Build GRCh38
rs532781136 Chr.13: 45151636-45151636 on Build GRCh38
rs754735108 Chr.13: 45151776-45151776 on Build GRCh38
rs897328348 Chr.13: 45151365-45151365 on Build GRCh38
rs567194890 Chr.13: 45151795-45151795 on Build GRCh38
rs1028026262 Chr.13: 45151370-45151370 on Build GRCh38
rs944109960 Chr.13: 45151635-45151635 on Build GRCh38
rs756528949 Chr.13: 45151769-45151769 on Build GRCh38
rs776462954 Chr.13: 45151817-45151817 on Build GRCh38
rs192539624 Chr.13: 45151502-45151502 on Build GRCh38
rs531729046 Chr.13: 45151546-45151546 on Build GRCh38
rs774053474 Chr.13: 45151644-45151644 on Build GRCh38
rs758869469 Chr.13: 45151639-45151639 on Build GRCh38
rs763206968 Chr.13: 45151715-45151715 on Build GRCh38
rs894821343 Chr.13: 45151570-45151570 on Build GRCh38
rs941071334 Chr.13: 45151616-45151616 on Build GRCh38
rs13378297 Chr.13: 45151628-45151628 on Build GRCh38
rs951896900 Chr.13: 45151392-45151392 on Build GRCh38
rs748925935 Chr.13: 45151775-45151775 on Build GRCh38
rs750701407 Chr.13: 45151768-45151768 on Build GRCh38
rs1017487933 Chr.13: 45151360-45151360 on Build GRCh38
rs995532690 Chr.13: 45151696-45151696 on Build GRCh38
rs1020408726 Chr.13: 45151753-45151753 on Build GRCh38
rs746742188 Chr.13: 45151802-45151802 on Build GRCh38
rs757703487 Chr.13: 45151730-45151730 on Build GRCh38
rs550537475 Chr.13: 45151552-45151552 on Build GRCh38
rs994338648 Chr.13: 45151389-45151392 on Build GRCh38
rs985455107 Chr.13: 45151433-45151433 on Build GRCh38
rs752948331 Chr.13: 45151703-45151703 on Build GRCh38
rs543020068 Chr.13: 45151491-45151491 on Build GRCh38

View Full Product Details