Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45192728-45193239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F2
Entrez Gene ID
2963
Gene Name
general transcription factor IIF subunit 2
Gene Aliases
BTF4, RAP30, TF2F2, TFIIF
UniGene
Hs.654582
Gene Chromosome Location
Chr.13: 45119830-45284105 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

Gene Symbol
KCTD4
Entrez Gene ID
386618
Gene Name
potassium channel tetramerization domain containing 4
Gene Aliases
bA321C24.3
UniGene
Hs.23406
Gene Chromosome Location
Chr.13: 45192853-45201040 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

SNP Detail

SNP ID Chromosome Location
rs41289545 Chr.13: 45192971-45192971 on Build GRCh38
rs1051457478 Chr.13: 45192814-45192814 on Build GRCh38
rs975810061 Chr.13: 45193161-45193161 on Build GRCh38
rs967503841 Chr.13: 45193181-45193181 on Build GRCh38
rs1025472087 Chr.13: 45193014-45193014 on Build GRCh38
rs981264444 Chr.13: 45193028-45193028 on Build GRCh38
rs769944740 Chr.13: 45193165-45193165 on Build GRCh38
rs892448000 Chr.13: 45192938-45192938 on Build GRCh38
rs558690905 Chr.13: 45192999-45192999 on Build GRCh38
rs984691177 Chr.13: 45193079-45193079 on Build GRCh38
rs1034635465 Chr.13: 45192944-45192944 on Build GRCh38
rs958860441 Chr.13: 45192957-45192957 on Build GRCh38
rs763757325 Chr.13: 45192855-45192855 on Build GRCh38
rs890205587 Chr.13: 45192843-45192843 on Build GRCh38
rs951870355 Chr.13: 45193033-45193033 on Build GRCh38
rs79891858 Chr.13: 45192926-45192926 on Build GRCh38
rs969464071 Chr.13: 45193018-45193018 on Build GRCh38
rs781246627 Chr.13: 45193030-45193032 on Build GRCh38
rs1050583353 Chr.13: 45192799-45192799 on Build GRCh38
rs898851495 Chr.13: 45192915-45192915 on Build GRCh38
rs907818065 Chr.13: 45193139-45193139 on Build GRCh38
rs563291492 Chr.13: 45193062-45193062 on Build GRCh38
rs757053332 Chr.13: 45193000-45193000 on Build GRCh38
rs922939168 Chr.13: 45193179-45193179 on Build GRCh38
rs932135329 Chr.13: 45192770-45192770 on Build GRCh38
rs1014611546 Chr.13: 45192900-45192900 on Build GRCh38
rs931712908 Chr.13: 45193180-45193180 on Build GRCh38
rs1004920198 Chr.13: 45192886-45192886 on Build GRCh38
rs554866513 Chr.13: 45193141-45193141 on Build GRCh38
rs931848110 Chr.13: 45192760-45192760 on Build GRCh38

View Full Product Details