Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45194301-45194811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F2
Entrez Gene ID
2963
Gene Name
general transcription factor IIF subunit 2
Gene Aliases
BTF4, RAP30, TF2F2, TFIIF
UniGene
Hs.654582
Gene Chromosome Location
Chr.13: 45119830-45284105 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

Gene Symbol
KCTD4
Entrez Gene ID
386618
Gene Name
potassium channel tetramerization domain containing 4
Gene Aliases
bA321C24.3
UniGene
Hs.23406
Gene Chromosome Location
Chr.13: 45192853-45201040 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4635915 c.43G>C p.E15Q Chr.13: 45194525-45194525 on Build GRCh38
4819662 c.8G>A p.R3H Chr.13: 45194560-45194560 on Build GRCh38
6692096 c.31G>T p.E11* Chr.13: 45194537-45194537 on Build GRCh38
6692097 c.237A>G p.I79M Chr.13: 45194331-45194331 on Build GRCh38
4575946 c.192T>G p.N64K Chr.13: 45194376-45194376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756414460 Chr.13: 45194468-45194468 on Build GRCh38
rs763027796 Chr.13: 45194545-45194545 on Build GRCh38
rs751433308 Chr.13: 45194551-45194551 on Build GRCh38
rs371658910 Chr.13: 45194358-45194358 on Build GRCh38
rs932860114 Chr.13: 45194687-45194687 on Build GRCh38
rs1031943507 Chr.13: 45194515-45194515 on Build GRCh38
rs901286870 Chr.13: 45194693-45194693 on Build GRCh38
rs757491963 Chr.13: 45194342-45194342 on Build GRCh38
rs200145636 Chr.13: 45194356-45194356 on Build GRCh38
rs775630555 Chr.13: 45194345-45194347 on Build GRCh38
rs527784178 Chr.13: 45194726-45194726 on Build GRCh38
rs755078847 Chr.13: 45194579-45194579 on Build GRCh38
rs369513732 Chr.13: 45194474-45194474 on Build GRCh38
rs1045976312 Chr.13: 45194425-45194425 on Build GRCh38
rs746630026 Chr.13: 45194507-45194507 on Build GRCh38
rs745561567 Chr.13: 45194748-45194748 on Build GRCh38
rs544470707 Chr.13: 45194532-45194532 on Build GRCh38
rs771462792 Chr.13: 45194643-45194643 on Build GRCh38
rs369829244 Chr.13: 45194607-45194607 on Build GRCh38
rs752486870 Chr.13: 45194448-45194448 on Build GRCh38
rs1008648305 Chr.13: 45194501-45194501 on Build GRCh38
rs962284670 Chr.13: 45194510-45194510 on Build GRCh38
rs148124193 Chr.13: 45194511-45194511 on Build GRCh38
rs539886664 Chr.13: 45194491-45194491 on Build GRCh38
rs748470801 Chr.13: 45194487-45194487 on Build GRCh38
rs765274069 Chr.13: 45194445-45194445 on Build GRCh38
rs541301933 Chr.13: 45194639-45194639 on Build GRCh38
rs778251048 Chr.13: 45194496-45194496 on Build GRCh38
rs1009225109 Chr.13: 45194738-45194738 on Build GRCh38
rs940032742 Chr.13: 45194652-45194652 on Build GRCh38
rs781583099 Chr.13: 45194348-45194348 on Build GRCh38
rs749741665 Chr.13: 45194367-45194367 on Build GRCh38
rs888972853 Chr.13: 45194703-45194703 on Build GRCh38
rs1018830599 Chr.13: 45194503-45194503 on Build GRCh38
rs760417060 Chr.13: 45194417-45194417 on Build GRCh38
rs777996346 Chr.13: 45194612-45194612 on Build GRCh38
rs200076066 Chr.13: 45194359-45194359 on Build GRCh38
rs1049761734 Chr.13: 45194699-45194699 on Build GRCh38
rs891219541 Chr.13: 45194709-45194709 on Build GRCh38
rs767811573 Chr.13: 45194457-45194457 on Build GRCh38
rs949732704 Chr.13: 45194413-45194413 on Build GRCh38
rs759387945 Chr.13: 45194442-45194442 on Build GRCh38
rs149350558 Chr.13: 45194454-45194454 on Build GRCh38
rs759102946 Chr.13: 45194528-45194528 on Build GRCh38
rs750585917 Chr.13: 45194462-45194462 on Build GRCh38
rs567650112 Chr.13: 45194783-45194783 on Build GRCh38
rs772880987 Chr.13: 45194406-45194406 on Build GRCh38
rs12877778 Chr.13: 45194642-45194642 on Build GRCh38
rs562563207 Chr.13: 45194560-45194560 on Build GRCh38
rs549439610 Chr.13: 45194764-45194764 on Build GRCh38
rs752607396 Chr.13: 45194332-45194332 on Build GRCh38
rs766614055 Chr.13: 45194711-45194712 on Build GRCh38
rs147004947 Chr.13: 45194472-45194472 on Build GRCh38
rs769214534 Chr.13: 45194382-45194382 on Build GRCh38
rs373141842 Chr.13: 45194567-45194567 on Build GRCh38
rs1016029864 Chr.13: 45194727-45194727 on Build GRCh38
rs949659534 Chr.13: 45194546-45194546 on Build GRCh38
rs1032248770 Chr.13: 45194711-45194711 on Build GRCh38
rs996874881 Chr.13: 45194461-45194461 on Build GRCh38
rs776263100 Chr.13: 45194438-45194438 on Build GRCh38
rs938207095 Chr.13: 45194631-45194631 on Build GRCh38
rs763965849 Chr.13: 45194547-45194547 on Build GRCh38
rs893452434 Chr.13: 45194728-45194728 on Build GRCh38
rs1004278141 Chr.13: 45194360-45194360 on Build GRCh38
rs753181440 Chr.13: 45194589-45194589 on Build GRCh38
rs141883616 Chr.13: 45194539-45194539 on Build GRCh38
rs893298529 Chr.13: 45194660-45194660 on Build GRCh38
rs1037100601 Chr.13: 45194678-45194678 on Build GRCh38
rs781071691 Chr.13: 45194383-45194383 on Build GRCh38
rs539182975 Chr.13: 45194350-45194350 on Build GRCh38
rs573630345 Chr.13: 45194526-45194526 on Build GRCh38
rs774836783 Chr.13: 45194742-45194742 on Build GRCh38
rs761142952 Chr.13: 45194531-45194531 on Build GRCh38
rs766636820 Chr.13: 45194561-45194561 on Build GRCh38
rs765785812 Chr.13: 45194434-45194434 on Build GRCh38
rs771661691 Chr.13: 45194393-45194393 on Build GRCh38
rs547072230 Chr.13: 45194697-45194697 on Build GRCh38
rs1008202478 Chr.13: 45194710-45194710 on Build GRCh38
rs9534059 Chr.13: 45194624-45194624 on Build GRCh38
rs915565506 Chr.13: 45194400-45194400 on Build GRCh38
rs748525948 Chr.13: 45194384-45194384 on Build GRCh38
rs376008425 Chr.13: 45194388-45194388 on Build GRCh38
rs116237860 Chr.13: 45194575-45194575 on Build GRCh38
rs1048394518 Chr.13: 45194353-45194353 on Build GRCh38

View Full Product Details