Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48256096-48256605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITM2B
Entrez Gene ID
9445
Gene Name
integral membrane protein 2B
Gene Aliases
ABRI, BRI, BRI2, BRICD2B, E25B, E3-16, FBD, RDGCA, imBRI2
UniGene
Hs.643683
Gene Chromosome Location
Chr.13: 48233138-48262096 on Build GRCh38
Transcripts
NM_021999.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3469140 c.331C>T p.L111F Chr.13: 48256261-48256261 on Build GRCh38
947774 c.341C>T p.T114I Chr.13: 48256271-48256271 on Build GRCh38
947775 c.349G>T p.E117* Chr.13: 48256279-48256279 on Build GRCh38
5027766 c.279C>A p.Y93* Chr.13: 48256209-48256209 on Build GRCh38
947776 c.382G>A p.E128K Chr.13: 48256312-48256312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764206598 Chr.13: 48256295-48256295 on Build GRCh38
rs767959003 Chr.13: 48256308-48256308 on Build GRCh38
rs768824125 Chr.13: 48256433-48256433 on Build GRCh38
rs373084833 Chr.13: 48256148-48256148 on Build GRCh38
rs775445893 Chr.13: 48256164-48256164 on Build GRCh38
rs764153376 Chr.13: 48256173-48256173 on Build GRCh38
rs905537288 Chr.13: 48256408-48256408 on Build GRCh38
rs370484810 Chr.13: 48256301-48256301 on Build GRCh38
rs983245766 Chr.13: 48256167-48256168 on Build GRCh38
rs758433423 Chr.13: 48256392-48256392 on Build GRCh38
rs774433204 Chr.13: 48256435-48256435 on Build GRCh38
rs74074156 Chr.13: 48256419-48256419 on Build GRCh38
rs763071123 Chr.13: 48256293-48256293 on Build GRCh38
rs779702752 Chr.13: 48256240-48256240 on Build GRCh38
rs770816243 Chr.13: 48256420-48256420 on Build GRCh38
rs755741361 Chr.13: 48256229-48256229 on Build GRCh38
rs912179699 Chr.13: 48256581-48256581 on Build GRCh38
rs1036021515 Chr.13: 48256432-48256432 on Build GRCh38
rs762336440 Chr.13: 48256307-48256307 on Build GRCh38
rs113956182 Chr.13: 48256214-48256214 on Build GRCh38
rs963480184 Chr.13: 48256471-48256471 on Build GRCh38
rs923341920 Chr.13: 48256449-48256449 on Build GRCh38
rs1014765709 Chr.13: 48256470-48256470 on Build GRCh38
rs935508599 Chr.13: 48256291-48256291 on Build GRCh38
rs542014399 Chr.13: 48256429-48256429 on Build GRCh38
rs779568121 Chr.13: 48256297-48256299 on Build GRCh38
rs775448316 Chr.13: 48256548-48256548 on Build GRCh38
rs867626030 Chr.13: 48256496-48256496 on Build GRCh38
rs541331841 Chr.13: 48256270-48256270 on Build GRCh38
rs968251360 Chr.13: 48256360-48256360 on Build GRCh38
rs200085682 Chr.13: 48256333-48256333 on Build GRCh38
rs751018650 Chr.13: 48256297-48256302 on Build GRCh38
rs746769961 Chr.13: 48256275-48256275 on Build GRCh38
rs749707917 Chr.13: 48256186-48256186 on Build GRCh38
rs754668102 Chr.13: 48256366-48256367 on Build GRCh38
rs3818455 Chr.13: 48256465-48256465 on Build GRCh38
rs1001814355 Chr.13: 48256523-48256523 on Build GRCh38
rs754657407 Chr.13: 48256347-48256347 on Build GRCh38
rs772051695 Chr.13: 48256135-48256135 on Build GRCh38
rs766650893 Chr.13: 48256338-48256338 on Build GRCh38
rs1038670809 Chr.13: 48256514-48256514 on Build GRCh38
rs771173753 Chr.13: 48256366-48256366 on Build GRCh38
rs757484295 Chr.13: 48256246-48256246 on Build GRCh38
rs142100294 Chr.13: 48256510-48256510 on Build GRCh38
rs200601547 Chr.13: 48256156-48256156 on Build GRCh38
rs185653495 Chr.13: 48256500-48256500 on Build GRCh38
rs199641696 Chr.13: 48256166-48256166 on Build GRCh38
rs749561578 Chr.13: 48256431-48256431 on Build GRCh38
rs752215006 Chr.13: 48256382-48256382 on Build GRCh38
rs9332284 Chr.13: 48256254-48256254 on Build GRCh38
rs750762827 Chr.13: 48256315-48256315 on Build GRCh38
rs775848254 Chr.13: 48256286-48256286 on Build GRCh38
rs748146945 Chr.13: 48256255-48256255 on Build GRCh38
rs896239855 Chr.13: 48256577-48256577 on Build GRCh38
rs201982776 Chr.13: 48256177-48256177 on Build GRCh38
rs9332285 Chr.13: 48256572-48256572 on Build GRCh38
rs528104609 Chr.13: 48256522-48256522 on Build GRCh38
rs377585277 Chr.13: 48256266-48256266 on Build GRCh38
rs1005398400 Chr.13: 48256513-48256513 on Build GRCh38
rs778609115 Chr.13: 48256348-48256348 on Build GRCh38
rs754586409 Chr.13: 48256253-48256253 on Build GRCh38
rs886908143 Chr.13: 48256502-48256502 on Build GRCh38
rs753588397 Chr.13: 48256343-48256343 on Build GRCh38
rs74074155 Chr.13: 48256185-48256185 on Build GRCh38
rs563897746 Chr.13: 48256521-48256521 on Build GRCh38
rs143931252 Chr.13: 48256183-48256183 on Build GRCh38

View Full Product Details