Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 96836242-96836763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS6ST3
Entrez Gene ID
266722
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
HS6ST-3
UniGene
Hs.171001
Gene Chromosome Location
Chr.13: 96090634-96839562 on Build GRCh38
Transcripts
NM_153456.3

SNP Detail

SNP ID Chromosome Location
rs187026524 Chr.13: 96836481-96836481 on Build GRCh38
rs569181596 Chr.13: 96836637-96836637 on Build GRCh38
rs371238389 Chr.13: 96836727-96836727 on Build GRCh38
rs373489181 Chr.13: 96836636-96836636 on Build GRCh38
rs907864034 Chr.13: 96836706-96836706 on Build GRCh38
rs761104877 Chr.13: 96836559-96836559 on Build GRCh38
rs954775912 Chr.13: 96836677-96836677 on Build GRCh38
rs530238019 Chr.13: 96836404-96836404 on Build GRCh38
rs754151388 Chr.13: 96836704-96836704 on Build GRCh38
rs1025402652 Chr.13: 96836560-96836564 on Build GRCh38
rs941985846 Chr.13: 96836720-96836720 on Build GRCh38
rs896491075 Chr.13: 96836385-96836385 on Build GRCh38
rs538041825 Chr.13: 96836306-96836306 on Build GRCh38
rs923321961 Chr.13: 96836288-96836288 on Build GRCh38
rs562328196 Chr.13: 96836339-96836339 on Build GRCh38
rs966960502 Chr.13: 96836599-96836599 on Build GRCh38
rs1012939097 Chr.13: 96836424-96836424 on Build GRCh38
rs545434345 Chr.13: 96836311-96836311 on Build GRCh38
rs951076303 Chr.13: 96836560-96836560 on Build GRCh38
rs995667506 Chr.13: 96836507-96836507 on Build GRCh38
rs41405545 Chr.13: 96836608-96836608 on Build GRCh38
rs772966245 Chr.13: 96836686-96836686 on Build GRCh38
rs144996110 Chr.13: 96836381-96836381 on Build GRCh38
rs968973435 Chr.13: 96836709-96836709 on Build GRCh38
rs983487298 Chr.13: 96836682-96836682 on Build GRCh38
rs374986000 Chr.13: 96836533-96836533 on Build GRCh38
rs142819147 Chr.13: 96836365-96836365 on Build GRCh38
rs746348985 Chr.13: 96836265-96836265 on Build GRCh38
rs975282872 Chr.13: 96836654-96836654 on Build GRCh38
rs113741873 Chr.13: 96836270-96836270 on Build GRCh38
rs939912291 Chr.13: 96836330-96836330 on Build GRCh38
rs138793793 Chr.13: 96836396-96836396 on Build GRCh38
rs887064359 Chr.13: 96836327-96836327 on Build GRCh38
rs1047723098 Chr.13: 96836316-96836316 on Build GRCh38
rs557059082 Chr.13: 96836269-96836269 on Build GRCh38
rs189698571 Chr.13: 96836510-96836510 on Build GRCh38
rs923385839 Chr.13: 96836671-96836671 on Build GRCh38
rs896185222 Chr.13: 96836491-96836491 on Build GRCh38
rs1045308676 Chr.13: 96836457-96836457 on Build GRCh38
rs554608508 Chr.13: 96836572-96836572 on Build GRCh38
rs1044210068 Chr.13: 96836291-96836291 on Build GRCh38
rs1038231187 Chr.13: 96836338-96836338 on Build GRCh38
rs539868601 Chr.13: 96836645-96836645 on Build GRCh38
rs1020568117 Chr.13: 96836573-96836573 on Build GRCh38

View Full Product Details