Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31292023-31292526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR5A
Entrez Gene ID
25938
Gene Name
HEAT repeat containing 5A
Gene Aliases
C14orf125
UniGene
Hs.744979
Gene Chromosome Location
Chr.14: 31291788-31420582 on Build GRCh38
Transcripts
NM_015473.3

SNP Detail

SNP ID Chromosome Location
rs547806641 Chr.14: 31292257-31292257 on Build GRCh38
rs1028112321 Chr.14: 31292131-31292131 on Build GRCh38
rs953007384 Chr.14: 31292474-31292474 on Build GRCh38
rs4981091 Chr.14: 31292340-31292340 on Build GRCh38
rs949658763 Chr.14: 31292053-31292053 on Build GRCh38
rs916735887 Chr.14: 31292326-31292326 on Build GRCh38
rs1008454902 Chr.14: 31292498-31292498 on Build GRCh38
rs954124699 Chr.14: 31292141-31292141 on Build GRCh38
rs899214921 Chr.14: 31292097-31292097 on Build GRCh38
rs888420134 Chr.14: 31292260-31292260 on Build GRCh38
rs897695844 Chr.14: 31292164-31292164 on Build GRCh38
rs899296605 Chr.14: 31292461-31292461 on Build GRCh38
rs944521075 Chr.14: 31292093-31292093 on Build GRCh38
rs575385791 Chr.14: 31292315-31292315 on Build GRCh38
rs977714870 Chr.14: 31292171-31292171 on Build GRCh38
rs1008135556 Chr.14: 31292145-31292145 on Build GRCh38
rs1037767439 Chr.14: 31292090-31292090 on Build GRCh38
rs1000373244 Chr.14: 31292469-31292469 on Build GRCh38
rs1049037176 Chr.14: 31292190-31292190 on Build GRCh38
rs1014376681 Chr.14: 31292369-31292369 on Build GRCh38
rs886426218 Chr.14: 31292080-31292080 on Build GRCh38
rs985872598 Chr.14: 31292065-31292065 on Build GRCh38
rs75443632 Chr.14: 31292425-31292425 on Build GRCh38
rs139330258 Chr.14: 31292113-31292113 on Build GRCh38
rs41508745 Chr.14: 31292279-31292279 on Build GRCh38
rs1037630297 Chr.14: 31292460-31292463 on Build GRCh38
rs749572846 Chr.14: 31292163-31292163 on Build GRCh38
rs530725699 Chr.14: 31292476-31292476 on Build GRCh38
rs1005261543 Chr.14: 31292088-31292088 on Build GRCh38
rs967544080 Chr.14: 31292450-31292450 on Build GRCh38
rs183005469 Chr.14: 31292443-31292443 on Build GRCh38
rs746010170 Chr.14: 31292069-31292069 on Build GRCh38
rs1019921507 Chr.14: 31292149-31292149 on Build GRCh38
rs895898680 Chr.14: 31292324-31292324 on Build GRCh38
rs1033240456 Chr.14: 31292471-31292471 on Build GRCh38
rs930528050 Chr.14: 31292173-31292173 on Build GRCh38

View Full Product Details