Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31292883-31293396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR5A
Entrez Gene ID
25938
Gene Name
HEAT repeat containing 5A
Gene Aliases
C14orf125
UniGene
Hs.744979
Gene Chromosome Location
Chr.14: 31291788-31420582 on Build GRCh38
Transcripts
NM_015473.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3495682 c.6111C>T p.T2037T Chr.14: 31293317-31293317 on Build GRCh38
3495684 c.6034G>A p.E2012K Chr.14: 31293394-31293394 on Build GRCh38
955276 c.6062C>T p.S2021F Chr.14: 31293366-31293366 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1023712956 Chr.14: 31293005-31293005 on Build GRCh38
rs761662602 Chr.14: 31293317-31293317 on Build GRCh38
rs371661207 Chr.14: 31293131-31293132 on Build GRCh38
rs375824559 Chr.14: 31293261-31293261 on Build GRCh38
rs943569289 Chr.14: 31293329-31293329 on Build GRCh38
rs759733714 Chr.14: 31293342-31293342 on Build GRCh38
rs1053719358 Chr.14: 31292933-31292933 on Build GRCh38
rs778011625 Chr.14: 31293349-31293349 on Build GRCh38
rs767766531 Chr.14: 31293343-31293343 on Build GRCh38
rs368350492 Chr.14: 31293207-31293207 on Build GRCh38
rs1026035348 Chr.14: 31292944-31292944 on Build GRCh38
rs762342261 Chr.14: 31293351-31293352 on Build GRCh38
rs755278426 Chr.14: 31293353-31293354 on Build GRCh38
rs373846019 Chr.14: 31293312-31293312 on Build GRCh38
rs776577748 Chr.14: 31293315-31293315 on Build GRCh38
rs951770744 Chr.14: 31292969-31292969 on Build GRCh38
rs200750007 Chr.14: 31293346-31293346 on Build GRCh38
rs56060018 Chr.14: 31293304-31293304 on Build GRCh38
rs1003692343 Chr.14: 31293024-31293024 on Build GRCh38
rs774884600 Chr.14: 31293294-31293295 on Build GRCh38
rs770415599 Chr.14: 31293260-31293260 on Build GRCh38
rs771904532 Chr.14: 31293294-31293294 on Build GRCh38
rs902334261 Chr.14: 31292915-31292915 on Build GRCh38
rs993784773 Chr.14: 31293355-31293355 on Build GRCh38
rs571547087 Chr.14: 31293321-31293321 on Build GRCh38
rs745509415 Chr.14: 31293264-31293264 on Build GRCh38
rs893685967 Chr.14: 31292942-31292942 on Build GRCh38
rs80303266 Chr.14: 31292929-31292929 on Build GRCh38
rs760585008 Chr.14: 31293307-31293307 on Build GRCh38
rs754304855 Chr.14: 31293356-31293356 on Build GRCh38
rs529392954 Chr.14: 31293102-31293106 on Build GRCh38
rs1054498003 Chr.14: 31293308-31293308 on Build GRCh38
rs147946608 Chr.14: 31293179-31293179 on Build GRCh38
rs193176387 Chr.14: 31293085-31293085 on Build GRCh38
rs780535466 Chr.14: 31293339-31293339 on Build GRCh38
rs977382179 Chr.14: 31293133-31293133 on Build GRCh38
rs534130020 Chr.14: 31293069-31293069 on Build GRCh38
rs777911336 Chr.14: 31293128-31293128 on Build GRCh38
rs751652730 Chr.14: 31293323-31293323 on Build GRCh38
rs369468079 Chr.14: 31293362-31293362 on Build GRCh38
rs1011920808 Chr.14: 31292981-31292981 on Build GRCh38
rs572944305 Chr.14: 31293051-31293051 on Build GRCh38
rs766479783 Chr.14: 31293318-31293318 on Build GRCh38
rs368151363 Chr.14: 31293295-31293296 on Build GRCh38
rs11850386 Chr.14: 31293122-31293122 on Build GRCh38
rs779954653 Chr.14: 31293368-31293368 on Build GRCh38
rs780873120 Chr.14: 31292986-31292986 on Build GRCh38
rs557981112 Chr.14: 31292998-31292998 on Build GRCh38
rs758507803 Chr.14: 31293176-31293176 on Build GRCh38
rs959266035 Chr.14: 31293096-31293096 on Build GRCh38
rs749175601 Chr.14: 31293132-31293132 on Build GRCh38
rs911862167 Chr.14: 31293102-31293111 on Build GRCh38
rs778469589 Chr.14: 31293367-31293367 on Build GRCh38
rs756419716 Chr.14: 31293347-31293347 on Build GRCh38
rs369998400 Chr.14: 31293300-31293300 on Build GRCh38
rs766887804 Chr.14: 31293106-31293107 on Build GRCh38
rs757723316 Chr.14: 31293295-31293295 on Build GRCh38
rs556476037 Chr.14: 31293276-31293277 on Build GRCh38
rs953623570 Chr.14: 31293101-31293102 on Build GRCh38

View Full Product Details