Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69658425-69658936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUSD6
Entrez Gene ID
9766
Gene Name
sushi domain containing 6
Gene Aliases
DRAGO, KIAA0247
UniGene
Hs.440025
Gene Chromosome Location
Chr.14: 69611593-69715144 on Build GRCh38
Transcripts
NM_014734.3 XM_011537412.1 XM_011537413.1 XM_017021838.1 XM_017021839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
88728 c.121+3_121+4insT p.? Chr.14: 69658716-69658717 on Build GRCh38
3815188 c.94A>C p.T32P Chr.14: 69658686-69658686 on Build GRCh38
6271609 c.58G>T p.G20* Chr.14: 69658650-69658650 on Build GRCh38
7192988 c.62A>G p.H21R Chr.14: 69658654-69658654 on Build GRCh38
321163 c.76C>T p.P26S Chr.14: 69658668-69658668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111720331 Chr.14: 69658913-69658913 on Build GRCh38
rs762224119 Chr.14: 69658583-69658583 on Build GRCh38
rs764731508 Chr.14: 69658643-69658643 on Build GRCh38
rs946581917 Chr.14: 69658829-69658829 on Build GRCh38
rs753389290 Chr.14: 69658809-69658809 on Build GRCh38
rs757494679 Chr.14: 69658564-69658564 on Build GRCh38
rs769155399 Chr.14: 69658564-69658564 on Build GRCh38
rs554931938 Chr.14: 69658788-69658788 on Build GRCh38
rs557802876 Chr.14: 69658689-69658689 on Build GRCh38
rs111343998 Chr.14: 69658714-69658714 on Build GRCh38
rs763846976 Chr.14: 69658651-69658651 on Build GRCh38
rs188576160 Chr.14: 69658524-69658524 on Build GRCh38
rs148135611 Chr.14: 69658603-69658603 on Build GRCh38
rs772456729 Chr.14: 69658614-69658614 on Build GRCh38
rs139836864 Chr.14: 69658707-69658707 on Build GRCh38
rs554798849 Chr.14: 69658720-69658720 on Build GRCh38
rs77651642 Chr.14: 69658552-69658552 on Build GRCh38
rs749431779 Chr.14: 69658685-69658685 on Build GRCh38
rs921624845 Chr.14: 69658778-69658778 on Build GRCh38
rs150672072 Chr.14: 69658632-69658632 on Build GRCh38
rs142465499 Chr.14: 69658700-69658700 on Build GRCh38
rs1037646963 Chr.14: 69658865-69658865 on Build GRCh38
rs993665894 Chr.14: 69658501-69658501 on Build GRCh38
rs774641535 Chr.14: 69658578-69658578 on Build GRCh38
rs974421590 Chr.14: 69658775-69658775 on Build GRCh38
rs368354387 Chr.14: 69658637-69658637 on Build GRCh38
rs866795980 Chr.14: 69658753-69658753 on Build GRCh38
rs1024843350 Chr.14: 69658530-69658530 on Build GRCh38
rs752306675 Chr.14: 69658646-69658646 on Build GRCh38
rs748186357 Chr.14: 69658752-69658755 on Build GRCh38
rs968515838 Chr.14: 69658886-69658886 on Build GRCh38
rs769608280 Chr.14: 69658903-69658903 on Build GRCh38
rs769245328 Chr.14: 69658713-69658713 on Build GRCh38
rs762490172 Chr.14: 69658647-69658647 on Build GRCh38
rs539268109 Chr.14: 69658673-69658673 on Build GRCh38
rs958227910 Chr.14: 69658764-69658764 on Build GRCh38
rs900793323 Chr.14: 69658750-69658750 on Build GRCh38
rs199606527 Chr.14: 69658573-69658573 on Build GRCh38
rs1030451132 Chr.14: 69658780-69658780 on Build GRCh38
rs201323176 Chr.14: 69658659-69658659 on Build GRCh38
rs779347533 Chr.14: 69658580-69658583 on Build GRCh38
rs763638918 Chr.14: 69658723-69658723 on Build GRCh38
rs1006364723 Chr.14: 69658748-69658748 on Build GRCh38
rs927179777 Chr.14: 69658904-69658904 on Build GRCh38
rs1030070069 Chr.14: 69658594-69658594 on Build GRCh38
rs966599643 Chr.14: 69658536-69658536 on Build GRCh38
rs375610588 Chr.14: 69658575-69658575 on Build GRCh38
rs751305170 Chr.14: 69658655-69658655 on Build GRCh38
rs1033731356 Chr.14: 69658496-69658496 on Build GRCh38
rs750333875 Chr.14: 69658749-69658749 on Build GRCh38
rs957687965 Chr.14: 69658570-69658570 on Build GRCh38
rs898469606 Chr.14: 69658471-69658471 on Build GRCh38
rs74060256 Chr.14: 69658728-69658728 on Build GRCh38
rs141925470 Chr.14: 69658625-69658625 on Build GRCh38
rs376306418 Chr.14: 69658670-69658670 on Build GRCh38
rs749548388 Chr.14: 69658584-69658584 on Build GRCh38
rs762280809 Chr.14: 69658611-69658611 on Build GRCh38
rs925199022 Chr.14: 69658566-69658566 on Build GRCh38
rs964325263 Chr.14: 69658576-69658576 on Build GRCh38
rs991986126 Chr.14: 69658526-69658526 on Build GRCh38
rs868766969 Chr.14: 69658810-69658810 on Build GRCh38
rs1032256510 Chr.14: 69658758-69658758 on Build GRCh38
rs774562375 Chr.14: 69658601-69658601 on Build GRCh38
rs754901096 Chr.14: 69658881-69658881 on Build GRCh38
rs769020564 Chr.14: 69658598-69658598 on Build GRCh38
rs139068073 Chr.14: 69658669-69658669 on Build GRCh38
rs530898402 Chr.14: 69658456-69658456 on Build GRCh38
rs770004623 Chr.14: 69658577-69658577 on Build GRCh38
rs145936627 Chr.14: 69658702-69658702 on Build GRCh38
rs761445736 Chr.14: 69658737-69658737 on Build GRCh38
rs767447818 Chr.14: 69658745-69658745 on Build GRCh38
rs1021386990 Chr.14: 69658884-69658884 on Build GRCh38
rs987151256 Chr.14: 69658804-69658804 on Build GRCh38
rs761201672 Chr.14: 69658712-69658712 on Build GRCh38
rs183138271 Chr.14: 69658493-69658493 on Build GRCh38
rs746085523 Chr.14: 69658580-69658580 on Build GRCh38

View Full Product Details