Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69712410-69712909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUSD6
Entrez Gene ID
9766
Gene Name
sushi domain containing 6
Gene Aliases
DRAGO, KIAA0247
UniGene
Hs.440025
Gene Chromosome Location
Chr.14: 69611593-69715144 on Build GRCh38
Transcripts
NM_014734.3 XM_011537412.1 XM_011537413.1 XM_017021838.1 XM_017021839.1

SNP Detail

SNP ID Chromosome Location
rs913067895 Chr.14: 69712667-69712667 on Build GRCh38
rs1018005429 Chr.14: 69712480-69712480 on Build GRCh38
rs909189952 Chr.14: 69712560-69712560 on Build GRCh38
rs8006097 Chr.14: 69712537-69712537 on Build GRCh38
rs962466375 Chr.14: 69712833-69712833 on Build GRCh38
rs141091839 Chr.14: 69712625-69712625 on Build GRCh38
rs780485660 Chr.14: 69712699-69712699 on Build GRCh38
rs151092088 Chr.14: 69712514-69712514 on Build GRCh38
rs1030577304 Chr.14: 69712509-69712509 on Build GRCh38
rs995212828 Chr.14: 69712869-69712869 on Build GRCh38
rs575382493 Chr.14: 69712669-69712669 on Build GRCh38
rs754130182 Chr.14: 69712700-69712700 on Build GRCh38
rs749324918 Chr.14: 69712709-69712709 on Build GRCh38
rs997955363 Chr.14: 69712711-69712711 on Build GRCh38
rs554702099 Chr.14: 69712726-69712726 on Build GRCh38
rs1006565240 Chr.14: 69712453-69712454 on Build GRCh38
rs1005109090 Chr.14: 69712764-69712764 on Build GRCh38
rs980138731 Chr.14: 69712816-69712816 on Build GRCh38
rs535039067 Chr.14: 69712452-69712452 on Build GRCh38
rs960358795 Chr.14: 69712862-69712865 on Build GRCh38
rs945930298 Chr.14: 69712677-69712677 on Build GRCh38
rs1034362078 Chr.14: 69712823-69712823 on Build GRCh38
rs867795317 Chr.14: 69712691-69712691 on Build GRCh38
rs751444255 Chr.14: 69712624-69712624 on Build GRCh38
rs553641626 Chr.14: 69712500-69712500 on Build GRCh38
rs557027956 Chr.14: 69712521-69712521 on Build GRCh38
rs993035831 Chr.14: 69712659-69712659 on Build GRCh38
rs567925669 Chr.14: 69712439-69712439 on Build GRCh38
rs1015303512 Chr.14: 69712824-69712824 on Build GRCh38
rs927393431 Chr.14: 69712629-69712629 on Build GRCh38
rs904181382 Chr.14: 69712693-69712693 on Build GRCh38
rs1016554206 Chr.14: 69712780-69712780 on Build GRCh38
rs778886248 Chr.14: 69712873-69712873 on Build GRCh38
rs1030879541 Chr.14: 69712714-69712714 on Build GRCh38
rs542592362 Chr.14: 69712707-69712707 on Build GRCh38
rs573205731 Chr.14: 69712737-69712737 on Build GRCh38
rs938520111 Chr.14: 69712633-69712633 on Build GRCh38
rs904215489 Chr.14: 69712436-69712436 on Build GRCh38
rs992944900 Chr.14: 69712877-69712878 on Build GRCh38
rs963356847 Chr.14: 69712800-69712800 on Build GRCh38
rs950999976 Chr.14: 69712511-69712511 on Build GRCh38
rs186973836 Chr.14: 69712508-69712508 on Build GRCh38
rs540940100 Chr.14: 69712842-69712842 on Build GRCh38
rs756403613 Chr.14: 69712664-69712664 on Build GRCh38
rs1003794839 Chr.14: 69712806-69712806 on Build GRCh38

View Full Product Details