Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69713575-69714086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUSD6
Entrez Gene ID
9766
Gene Name
sushi domain containing 6
Gene Aliases
DRAGO, KIAA0247
UniGene
Hs.440025
Gene Chromosome Location
Chr.14: 69611593-69715144 on Build GRCh38
Transcripts
NM_014734.3 XM_011537412.1 XM_011537413.1 XM_017021838.1 XM_017021839.1

SNP Detail

SNP ID Chromosome Location
rs1040771353 Chr.14: 69713939-69713939 on Build GRCh38
rs376006954 Chr.14: 69713763-69713763 on Build GRCh38
rs943814246 Chr.14: 69713823-69713823 on Build GRCh38
rs771932771 Chr.14: 69713812-69713815 on Build GRCh38
rs370416726 Chr.14: 69713823-69713832 on Build GRCh38
rs903181142 Chr.14: 69714051-69714051 on Build GRCh38
rs982503597 Chr.14: 69713670-69713670 on Build GRCh38
rs893762698 Chr.14: 69713971-69713971 on Build GRCh38
rs77747124 Chr.14: 69713705-69713705 on Build GRCh38
rs970069741 Chr.14: 69714017-69714017 on Build GRCh38
rs926397097 Chr.14: 69713924-69713924 on Build GRCh38
rs4902748 Chr.14: 69713836-69713836 on Build GRCh38
rs902365219 Chr.14: 69713949-69713949 on Build GRCh38
rs1012161247 Chr.14: 69713994-69713994 on Build GRCh38
rs3742906 Chr.14: 69713961-69713961 on Build GRCh38
rs571828359 Chr.14: 69713732-69713732 on Build GRCh38
rs533937215 Chr.14: 69713962-69713962 on Build GRCh38
rs190162174 Chr.14: 69713999-69713999 on Build GRCh38
rs533230237 Chr.14: 69713655-69713658 on Build GRCh38
rs1275817 Chr.14: 69714061-69714061 on Build GRCh38
rs1054467126 Chr.14: 69714062-69714062 on Build GRCh38
rs532702349 Chr.14: 69713748-69713748 on Build GRCh38
rs963828174 Chr.14: 69713827-69713827 on Build GRCh38
rs972964232 Chr.14: 69713780-69713780 on Build GRCh38
rs371221032 Chr.14: 69713969-69713969 on Build GRCh38
rs973800270 Chr.14: 69713873-69713873 on Build GRCh38
rs1033088222 Chr.14: 69713825-69713830 on Build GRCh38
rs568260401 Chr.14: 69713837-69713837 on Build GRCh38
rs113082039 Chr.14: 69713836-69713836 on Build GRCh38
rs1015239389 Chr.14: 69713862-69713862 on Build GRCh38
rs61982061 Chr.14: 69713695-69713695 on Build GRCh38
rs963860992 Chr.14: 69713614-69713614 on Build GRCh38
rs542327343 Chr.14: 69713686-69713686 on Build GRCh38
rs569081792 Chr.14: 69713785-69713785 on Build GRCh38
rs962399452 Chr.14: 69713863-69713864 on Build GRCh38
rs920146774 Chr.14: 69713790-69713790 on Build GRCh38
rs867874451 Chr.14: 69713948-69713948 on Build GRCh38
rs1035699280 Chr.14: 69714035-69714035 on Build GRCh38
rs931560093 Chr.14: 69713802-69713802 on Build GRCh38
rs961534765 Chr.14: 69714047-69714047 on Build GRCh38
rs546624603 Chr.14: 69713718-69713718 on Build GRCh38
rs1000211392 Chr.14: 69713822-69713822 on Build GRCh38
rs530738452 Chr.14: 69713661-69713661 on Build GRCh38
rs548353027 Chr.14: 69713813-69713813 on Build GRCh38
rs955285646 Chr.14: 69713844-69713844 on Build GRCh38
rs753355742 Chr.14: 69713836-69713837 on Build GRCh38
rs975722829 Chr.14: 69713649-69713649 on Build GRCh38
rs919309105 Chr.14: 69713819-69713819 on Build GRCh38
rs11538023 Chr.14: 69713815-69713815 on Build GRCh38
rs368819471 Chr.14: 69713837-69713838 on Build GRCh38
rs1003381669 Chr.14: 69714034-69714034 on Build GRCh38
rs928822804 Chr.14: 69713731-69713731 on Build GRCh38
rs972912025 Chr.14: 69714049-69714049 on Build GRCh38
rs118174170 Chr.14: 69713611-69713611 on Build GRCh38
rs1029986564 Chr.14: 69713843-69713843 on Build GRCh38
rs566761468 Chr.14: 69713808-69713808 on Build GRCh38
rs996731133 Chr.14: 69713834-69713834 on Build GRCh38
rs1056444969 Chr.14: 69713728-69713728 on Build GRCh38
rs937545617 Chr.14: 69713726-69713726 on Build GRCh38
rs764196882 Chr.14: 69713645-69713645 on Build GRCh38
rs779025583 Chr.14: 69713965-69713965 on Build GRCh38

View Full Product Details