Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88182990-88183488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK10
Entrez Gene ID
54207
Gene Name
potassium two pore domain channel subfamily K member 10
Gene Aliases
K2p10.1, PPP1R97, TREK-2, TREK2
UniGene
Hs.560255
Gene Chromosome Location
Chr.14: 88180108-88326912 on Build GRCh38
Transcripts
NM_021161.4 NM_138317.2 NM_138318.2 XM_011536840.2

SNP Detail

SNP ID Chromosome Location
rs969138243 Chr.14: 88183395-88183395 on Build GRCh38
rs773450011 Chr.14: 88183056-88183056 on Build GRCh38
rs973746920 Chr.14: 88183461-88183461 on Build GRCh38
rs953057479 Chr.14: 88183296-88183296 on Build GRCh38
rs887242581 Chr.14: 88183276-88183276 on Build GRCh38
rs1006485107 Chr.14: 88183246-88183246 on Build GRCh38
rs531505816 Chr.14: 88183424-88183424 on Build GRCh38
rs1029997537 Chr.14: 88183294-88183294 on Build GRCh38
rs925546758 Chr.14: 88183309-88183309 on Build GRCh38
rs1003039279 Chr.14: 88183325-88183325 on Build GRCh38
rs936862309 Chr.14: 88183136-88183136 on Build GRCh38
rs1006018448 Chr.14: 88183450-88183450 on Build GRCh38
rs933163950 Chr.14: 88183110-88183110 on Build GRCh38
rs553183756 Chr.14: 88183409-88183409 on Build GRCh38
rs887704202 Chr.14: 88183421-88183421 on Build GRCh38
rs556254362 Chr.14: 88183092-88183092 on Build GRCh38
rs137983850 Chr.14: 88183347-88183347 on Build GRCh38
rs765675777 Chr.14: 88183203-88183203 on Build GRCh38
rs1022071305 Chr.14: 88183051-88183051 on Build GRCh38
rs892566067 Chr.14: 88183146-88183146 on Build GRCh38
rs379643 Chr.14: 88183410-88183410 on Build GRCh38
rs1014646979 Chr.14: 88183267-88183267 on Build GRCh38
rs977502142 Chr.14: 88183403-88183403 on Build GRCh38
rs576822275 Chr.14: 88183126-88183126 on Build GRCh38
rs991478622 Chr.14: 88183019-88183019 on Build GRCh38
rs1013196336 Chr.14: 88183357-88183357 on Build GRCh38
rs1035893147 Chr.14: 88183346-88183346 on Build GRCh38
rs943271211 Chr.14: 88183458-88183458 on Build GRCh38
rs753985083 Chr.14: 88183169-88183169 on Build GRCh38
rs938042127 Chr.14: 88183412-88183412 on Build GRCh38
rs929041918 Chr.14: 88183207-88183207 on Build GRCh38
rs567587914 Chr.14: 88183049-88183049 on Build GRCh38
rs539990169 Chr.14: 88183300-88183300 on Build GRCh38
rs949386428 Chr.14: 88183157-88183157 on Build GRCh38
rs778040066 Chr.14: 88183307-88183307 on Build GRCh38
rs936832653 Chr.14: 88183312-88183312 on Build GRCh38
rs753166191 Chr.14: 88183239-88183239 on Build GRCh38
rs923482671 Chr.14: 88183065-88183065 on Build GRCh38
rs142556656 Chr.14: 88183416-88183416 on Build GRCh38
rs988731201 Chr.14: 88183330-88183330 on Build GRCh38
rs61975808 Chr.14: 88183142-88183142 on Build GRCh38
rs986419202 Chr.14: 88183102-88183102 on Build GRCh38
rs1046391448 Chr.14: 88183163-88183163 on Build GRCh38
rs112361264 Chr.14: 88183198-88183198 on Build GRCh38
rs962107925 Chr.14: 88183278-88183278 on Build GRCh38
rs559875475 Chr.14: 88183383-88183383 on Build GRCh38
rs572671387 Chr.14: 88183266-88183266 on Build GRCh38
rs378522 Chr.14: 88183066-88183066 on Build GRCh38
rs975733548 Chr.14: 88183281-88183281 on Build GRCh38
rs910415682 Chr.14: 88183455-88183455 on Build GRCh38
rs750070942 Chr.14: 88183331-88183331 on Build GRCh38

View Full Product Details