Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88183792-88184299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK10
Entrez Gene ID
54207
Gene Name
potassium two pore domain channel subfamily K member 10
Gene Aliases
K2p10.1, PPP1R97, TREK-2, TREK2
UniGene
Hs.560255
Gene Chromosome Location
Chr.14: 88180108-88326912 on Build GRCh38
Transcripts
NM_021161.4 NM_138317.2 NM_138318.2 XM_011536840.2

SNP Detail

SNP ID Chromosome Location
rs893575889 Chr.14: 88184264-88184264 on Build GRCh38
rs961650125 Chr.14: 88184205-88184205 on Build GRCh38
rs1054892150 Chr.14: 88183857-88183857 on Build GRCh38
rs527563493 Chr.14: 88183877-88183877 on Build GRCh38
rs746015982 Chr.14: 88183910-88183910 on Build GRCh38
rs1053313081 Chr.14: 88184182-88184182 on Build GRCh38
rs182570286 Chr.14: 88184239-88184239 on Build GRCh38
rs934884233 Chr.14: 88184161-88184161 on Build GRCh38
rs982263971 Chr.14: 88183893-88183893 on Build GRCh38
rs562192990 Chr.14: 88183820-88183820 on Build GRCh38
rs1033807062 Chr.14: 88184145-88184145 on Build GRCh38
rs1043271160 Chr.14: 88183869-88183869 on Build GRCh38
rs147900877 Chr.14: 88184236-88184236 on Build GRCh38
rs541043598 Chr.14: 88183932-88183932 on Build GRCh38
rs570104095 Chr.14: 88184014-88184014 on Build GRCh38
rs890415697 Chr.14: 88184198-88184198 on Build GRCh38
rs544624800 Chr.14: 88184273-88184273 on Build GRCh38
rs759564285 Chr.14: 88183894-88183894 on Build GRCh38
rs190372047 Chr.14: 88184015-88184015 on Build GRCh38
rs1007201291 Chr.14: 88183817-88183817 on Build GRCh38
rs568558821 Chr.14: 88184162-88184162 on Build GRCh38
rs534969309 Chr.14: 88184128-88184128 on Build GRCh38
rs1043604644 Chr.14: 88184140-88184140 on Build GRCh38
rs552579343 Chr.14: 88184021-88184021 on Build GRCh38
rs1010226487 Chr.14: 88183830-88183830 on Build GRCh38
rs972370856 Chr.14: 88183923-88183923 on Build GRCh38
rs144678478 Chr.14: 88184147-88184147 on Build GRCh38
rs566381645 Chr.14: 88184078-88184078 on Build GRCh38
rs948905098 Chr.14: 88183867-88183867 on Build GRCh38
rs1021236791 Chr.14: 88183862-88183862 on Build GRCh38
rs995069764 Chr.14: 88183940-88183940 on Build GRCh38
rs867232051 Chr.14: 88184091-88184091 on Build GRCh38
rs765332034 Chr.14: 88183896-88183896 on Build GRCh38
rs933113492 Chr.14: 88183933-88183933 on Build GRCh38
rs766077272 Chr.14: 88184081-88184081 on Build GRCh38
rs751939619 Chr.14: 88184007-88184007 on Build GRCh38
rs984596555 Chr.14: 88184160-88184160 on Build GRCh38
rs544935128 Chr.14: 88183875-88183875 on Build GRCh38
rs972770561 Chr.14: 88184211-88184211 on Build GRCh38
rs771074924 Chr.14: 88183824-88183824 on Build GRCh38
rs547584510 Chr.14: 88183895-88183895 on Build GRCh38
rs950697108 Chr.14: 88183819-88183819 on Build GRCh38
rs536387984 Chr.14: 88184176-88184176 on Build GRCh38
rs1039043944 Chr.14: 88184220-88184220 on Build GRCh38
rs886285536 Chr.14: 88184261-88184261 on Build GRCh38
rs943098934 Chr.14: 88183984-88183984 on Build GRCh38
rs906800665 Chr.14: 88184106-88184106 on Build GRCh38
rs140715033 Chr.14: 88183972-88183972 on Build GRCh38
rs910381980 Chr.14: 88184238-88184238 on Build GRCh38
rs753571808 Chr.14: 88184096-88184096 on Build GRCh38
rs114387190 Chr.14: 88183947-88183947 on Build GRCh38
rs927583088 Chr.14: 88183983-88183983 on Build GRCh38
rs919143049 Chr.14: 88184212-88184212 on Build GRCh38
rs749843696 Chr.14: 88184055-88184055 on Build GRCh38
rs1008909596 Chr.14: 88183913-88183913 on Build GRCh38
rs764508319 Chr.14: 88183981-88183981 on Build GRCh38
rs185960468 Chr.14: 88183876-88183876 on Build GRCh38
rs1053575672 Chr.14: 88184004-88184004 on Build GRCh38
rs144217248 Chr.14: 88183872-88183872 on Build GRCh38
rs1053928516 Chr.14: 88184242-88184242 on Build GRCh38
rs909864376 Chr.14: 88183921-88183921 on Build GRCh38
rs560876626 Chr.14: 88183922-88183922 on Build GRCh38
rs933228175 Chr.14: 88184221-88184221 on Build GRCh38

View Full Product Details